N3279
Monoclonal Anti-Nerve Growth Factor-β antibody produced in mouse
clone 25623, purified immunoglobulin, lyophilized powder
Synonym(s):
Anti-NGF-β
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
25623, monoclonal
form
lyophilized powder
species reactivity
human, rat, mouse
technique(s)
capture ELISA: 2-8 μg/mL (detection antibody at 0.1-0.4 μg/ml)
neutralization: 0.1-0.4 μg/mL
impurities
endotoxin, tested
isotype
IgG1
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... NGF(4803)
mouse ... Ngf(18049)
Immunogen
recombinant human NGF-β.
Biochem/physiol Actions
Nerve Growth factor (NGF) is a growth factor that is essential for the development and maintenance of the nervous system. NGF is composed of three non-covalently linked α, β and γ subunits. The β subunit is responsible for the biological functions of NGF. Nerve growth factor (NGF) binds to either to NGF receptor (NGFR) or the Trk receptor and can elicit biological responses through each of the receptors independently. NGF is important in the survival and differentiation of neurons and neural crest-derived cells during embryonic and early postnatal development and maintenance of peripheral neurons. NGF is also implicated in inflammation and allergic reactions.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.
Preparation Note
Purified using Protein G.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Josemberg S Baptista et al.
PloS one, 15(1), e0228155-e0228155 (2020-01-28)
Intervertebral disc (IVD) degeneration is a remodeling process mediated by several growth factors and cytokines. This process has been extensively studied in vitro and with pathologic specimens obtained during surgery for scoliosis or back pain. However, the occurrence and temporal
Seong Hoon Seo et al.
The Journal of investigative dermatology, 140(11), 2199-2209 (2020-04-06)
Itch in atopic dermatitis (AD) is aggravated under warm conditions. Transient receptor potential vanilloid (TRPV) 3, a member of the thermosensitive transient receptor potential channels, is activated by innocuous heat and is abundantly expressed in keratinocytes. The potential role of
Wolfgang Andreas Nockher et al.
The Journal of allergy and clinical immunology, 117(3), 583-589 (2006-03-09)
Understanding the complex pathophysiology of allergic diseases has been a main challenge of clinical and experimental research for many years. It is well known that the allergic inflammation triggers neuronal dysfunction and structural changes in the diseased tissues such as
C D Mills et al.
European journal of pain (London, England), 17(4), 469-479 (2012-08-24)
Injection of nerve growth factor (NGF) produces mechanical and thermal hypersensitivity in rodents and humans. Treatment with sequestering antibodies demonstrates the importance of NGF in various pain states, with efficacy seen in a number of animal pain models and in
Hrishikesh M Mehta et al.
Journal of neuroscience research, 90(12), 2259-2271 (2012-08-21)
Nerve growth factor (NGF) is a homodimer that binds to two distinct receptor types, TrkA and p75, to support survival and differentiation of neurons. The high-affinity binding on the cell surface is believed to involve a heteroreceptor complex, but its
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service