Skip to Content
Merck
All Photos(1)

Key Documents

N6009

Sigma-Aldrich

Nerve Growth Factor-2.5S from murine submaxillary gland

lyophilized powder, suitable for cell culture

Synonym(s):

NGF-2.5S

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Nerve Growth Factor-2.5S from murine submaxillary gland, NGF-2.5S, lyophilized powder, suitable for cell culture

biological source

mouse

Quality Level

form

lyophilized powder

potency

0.1-30 ng/mL EC50 (using PC-12 cells)

mol wt

26.5 kDa

packaging

pkg of 4X25 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Ngf(18049)

General description

Nerve growth factor (NGF) is produced by different cell types: structural, accessory, and immune cells. It belongs to the neurotrophin family of trophic factors.

Application

Nerve Growth Factor-2.5S from murine submaxillary gland has been used:
  • in Ham′s F-12 medium for performing growth cone collapse assay using dorsal root ganglions (DRGs)
  • as a media supplement in the “process compartment” for microfluidic chamber studies
  • to treat mid-luteal cells to study its effect on choline acetyltransferase (CHAT) expression

Biochem/physiol Actions

Nerve growth factor (NGF) is neurotrophic and maintains sympathetic and sensory neurons of the peripheral nervous system and forebrain cholinergic neurons. NGF promotes the development of neurite-like filaments from chick embryo dorsal root ganglia and rat PC12 pheochromocytoma cells. NGF may have a role in in vivo fetal development and nerve regeneration especially, after ischemic, surgical, or chemical injuries. NGF may also be associated with the central nervous system. It exhibits ameliorating effects against ulcers (human cutaneous, corneal ulcers, and pressure ulcers), glaucoma, maculopathy, and retinitis pigmentosa. NGF-2.5S is a type of NGF-b originally derived under dissociative conditions and often varies from NGF-b due to proteolysis arising during its purification.

Other Notes

2.5S subunit of nerve growth factor-7S (NGF-7S) is essentially the β-subunit when isolated from male mouse submaxillary glands under initially dissociative conditions by a modification of the method of Bocchini and Angeletti.

Physical form

Lyophilized from a 0.2 μm filtered solution in sodium acetate buffer.

Analysis Note

The biological activity is measured using a 3-day MTT assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nessma Sultan et al.
Neural regeneration research, 16(9), 1821-1828 (2021-01-30)
Dental pulp stem cells (DPSCs) secrete neurotrophic factors which may play an important therapeutic role in neural development, maintenance and repair. To test this hypothesis, DPSCs-conditioned medium (DPSCs-CM) was collected from 72 hours serum-free DPSCs cultures. The impact of DPSCs-derived
Masoud Shekarabi et al.
Methods in molecular biology (Clifton, N.J.), 2311, 177-184 (2021-05-26)
Preparations of peripheral sensory neurons from rodents are essential for studying the molecular mechanism of neuronal survival and physiology. Although, isolating and culturing these neurons proves difficult, often these preparations are contaminated with nonneuronal proliferating cells. Here, we describe an
Hongyue Dai et al.
FASEB bioAdvances, 4(1), 76-89 (2022-01-14)
Tumor innervation has recently been documented and characterized in various settings and tumor types. However, the role that nerves innervating tumors play in the pathogenesis of cancer has not been clarified. In this study, we searched for neural signaling from
Haeman Jang et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(33), 14063-14068 (2009-08-12)
One of the greatest influenza pandemic threats at this time is posed by the highly pathogenic H5N1 avian influenza viruses. To date, 61% of the 433 known human cases of H5N1 infection have proved fatal. Animals infected by H5N1 viruses
Hidetada Matsuoka et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 68(10), 679-690 (2020-09-05)
TWIK-related acid-sensitive K+ (TASK) homomeric channels, TASK1 and TASK3, are present in PC12 cells. The channels do not heteromerize due plausibly to a lack of p11 protein. Single-channel recording reveals that most of the rat carotid body (CB) glomus cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service