Skip to Content
Merck
All Photos(1)

Key Documents

N2630

Sigma-Aldrich

Nicotinamide 1,N6-ethenoadenine dinucleotide

≥98%

Synonym(s):

ε-NAD, 1,N6-Etheno NAD, 1,N6-Ethenonicotinamide adenine dinucleotide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27N7O14P2
CAS Number:
Molecular Weight:
687.45
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (Organic)

Assay

≥98%

form

powder

solubility

H2O: soluble-250 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

NC(=O)C1=CC=C[N](=C1)C2OC(COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)n4cnc5c4ncn6ccnc56)C(O)C2O

InChI

1S/C23H28N7O14P2/c24-19(35)11-2-1-4-28(6-11)22-17(33)15(31)12(42-22)7-40-45(36,37)44-46(38,39)41-8-13-16(32)18(34)23(43-13)30-10-26-14-20-25-3-5-29(20)9-27-21(14)30/h1-6,9-10,12-13,15-18,22-23,31-34H,7-8H2,(H2,24,35)(H,36,37)(H,38,39)

InChI key

AVLUSNAXLARRGJ-UHFFFAOYSA-N

Related Categories

Application

Nicotinamide 1,N6-ethenoadenine dinucleotide (ε-NAD) is an NAD fluorescent analog (fluorophore) that may be used to study ADP-ribosylation reactions and the structure/kinetics of NAD+ binding enzymes by assays such as etheno-NAD+ assay.

Other Notes

Analog of β-NAD

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V N Hingorani et al.
The Journal of biological chemistry, 263(36), 19804-19808 (1988-12-25)
Nicotinamide 1,N6-ethenoadenine dinucleotide (etheno-NAD, epsilon-NAD), a fluorescent analogue of NAD, was able to serve as a substrate for the bacterial toxin-catalyzed epsilon-ADP ribosylation of signal-transducing G-proteins. Pertussis toxin and transducin were used as a model system to characterize this reaction.
Y I Henis et al.
European journal of biochemistry, 112(1), 59-73 (1980-11-01)
The binding of nicotinamide--adenine dinucleotide (NAD+), nicotinamide--1,N6-ethenoadenine dinucleotide (epsilon NAD+), acetylpyridine--adenine dinucleotide (AcPyAD+), ATP, and adenosine diphosphoribose (ADP-ribose) to rabbit muscle glyceraldehyde-3-phosphate dehydrogenase (the enzyme) was examined. NAD+ and epsilon NAD+ were found to bind to the apoenzyme in a
D Jorcke et al.
Advances in experimental medicine and biology, 419, 447-451 (1997-01-01)
Treatment of isolated bovine liver mitochondria with either detergents or a crude pancreatic lipase, steapsin, resulted in solubilization of NAD+ glycohydrolase (NADase) activity. The two forms of this enzyme can be visualized directly in SDS-polyacrylamide gels (PAGs) by a fluorescence
R G Lowery et al.
The Journal of biological chemistry, 263(32), 16714-16719 (1988-11-15)
The enzyme that catalyzes the ADP-ribosylation and concomitant inactivation of dinitrogenase reductase in Rhodospirillum rubrum has been purified greater than 19,000-fold to near homogeneity. We propose dinitrogenase reductase ADP-ribosyltransferase (DRAT) as the working name for the enzyme. DRAT activity is
James N Wilson et al.
Organic & biomolecular chemistry, 4(23), 4265-4274 (2006-11-15)
We describe the design, synthesis, and properties of nucleoside monomers in which the DNA base is replaced by fluorescent hydrocarbons and heterocycles, and the assembly of these monomers into DNA-like molecules in which the all bases are fluorescent. As monomers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service