Skip to Content
Merck
All Photos(1)

Key Documents

N216

Sigma-Aldrich

Monoclonal Anti-Na+/Ca2+ Exchanger antibody produced in mouse

clone C2C12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody product type

primary antibodies

clone

C2C12, monoclonal

form

liquid

mol wt

antigen 120 kDa (mature exchanger)
antigen 160 kDa (non-reduced)
antigen 70 kDa (proteolytic fragment)

species reactivity

human, rat, canine, rabbit, guinea pig

technique(s)

immunohistochemistry (frozen sections): 1:200
immunoprecipitation (IP): suitable
western blot: 1:1,000

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SLC8A1(6546)

General description

Na+/Ca2+ exchanger is also known as solute carrier family 8 member A1 (SLC8A1) and is present in the plasma membrane. It is predicted to have nine transmembrane segments and the gene encoding SLC8A1 has three promoters.

Specificity

The antibody reacts specifically with Na+/Ca2+ exchanger in kidney and cardiac tissues. By immunohistology, staining of the plasma membrane and intense staining of the cardiac T-tubular membrane is observed. The antibody is not recommended for immunoblotting of rat tissues.

Immunogen

dog cardiac Na+/Ca2+ exchanger. The epitope is in the region of amino acids 371-525, which is on the intracellular side of the plasma membrane.

Biochem/physiol Actions

Na+/Ca2+ exchanger (SLC8A1, solute carrier family 8 member A1) is involved in Ca2+ homeostasis. Depending on physiological conditions prevailing in the cell, it can either pump out Na+ or Ca2+.

Physical form

Solution in phosphate buffered saline, 1 mg/ml BSA containing 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wakam Chang et al.
Nucleus (Austin, Tex.), 6(1), 77-88 (2015-01-15)
Myoblast migration is essential for muscle development and repair; however, the factors that contribute to the polarity of migrating myoblasts are relatively unknown. We find that randomly migrating C2C12 myoblasts orient their centrosomes in the direction of migration. Using wounded
Sandhya Sriram et al.
Aging cell, 10(6), 931-948 (2011-07-21)
Abnormal levels of reactive oxygen species (ROS) and inflammatory cytokines have been observed in the skeletal muscle during muscle wasting including sarcopenia. However, the mechanisms that signal ROS production and prolonged maintenance of ROS levels during muscle wasting are not
Florian Steinberg et al.
The Journal of biological chemistry, 285(3), 2193-2202 (2009-11-19)
FGFRL1 (fibroblast growth factor receptor like 1) is the fifth and most recently discovered member of the fibroblast growth factor receptor (FGFR) family. With up to 50% amino acid similarity, its extracellular domain closely resembles that of the four conventional
Hanxing Wan et al.
Oncogene, 41(35), 4169-4182 (2022-07-27)
Plasma membrane Na+/Ca2+ exchanger 1 (NCX1) is a bidirectional ion transporter to operate in Ca2+ entry or exit modes, and TRPC1 is Ca2+-permeable channel. Both NCX1 and TRPC1 play critical roles in maintaining cytosolic free Ca2+ ([Ca2+]cyt) homeostasis in mammalian
Julian Vallejo et al.
PloS one, 11(3), e0150066-e0150066 (2016-03-10)
There is growing evidence that severe decline of skeletal muscle mass and function with age may be mitigated by exercise and dietary supplementation with protein and amino acid ingredient technologies. The purposes of this study were to examine the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service