Skip to Content
Merck
All Photos(3)

Documents

N0252

Sigma-Aldrich

4-Nitrophenyl decanoate

lipase substrate, ≥98% (TLC), powder

Synonym(s):

4-Nitrophenyl caprate, Decanoic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23NO4
CAS Number:
Molecular Weight:
293.36
Beilstein:
1915230
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

4-Nitrophenyl decanoate, lipase substrate

Assay

≥98% (TLC)

form

powder

solubility

chloroform: soluble 100 mg/mL, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C16H23NO4/c1-2-3-4-5-6-7-8-9-16(18)21-15-12-10-14(11-13-15)17(19)20/h10-13H,2-9H2,1H3

InChI key

RRNKCSIEGPOIKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl decanoate has been used as a substrate for assessing the lipase activity spectrophotometrically. It has also been used as a substrate for estimating the carboxylesterases activity.
4-Nitrophenyl decanoate has been used as a substrate to assess the carboxylesterase production by recombinant Penicillium griseoroseum T55 strain. It has also been used for general esterase activity assay of the hepatopancreas extract.

Biochem/physiol Actions

4-Nitrophenyl decanoate is a substrate for lipase enzyme. The extracellular lipase has greater hydrolysis reaction velocity with 4-nitrophenyl decanoate when compared with other 4-nitropheny esters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and characterization of an enzyme from the Greenshell? mussel Perna canaliculus that hydrolyses pectenotoxins and esters of okadaic acid
MacKenzie LA, et al.
Toxicon, 60(3), 406-419 (2012)
V M Gumerov et al.
Prikladnaia biokhimiia i mikrobiologiia, 48(4), 376-382 (2012-10-06)
As a result of sequencing the genome of the termophilic alkali-tolerant lipolytic bacterium Thermosyntropha lipolytica, the gene encoding a lipase secreted into the medium was identified. The recombinant enzyme was expressed in Escherichia coli. It was isolated, purified, and functionally
Methods in Molecular biology: Platelets and Megakaryocytes, volume 1, functional assays, 27-27 (2012)
Janaina Marques de Almeida et al.
Scientific reports, 8(1), 10000-10000 (2018-07-04)
We determined the effect of the His-tag on the structure, activity, stability and immobilization of LipC12, a highly active lipase from a metagenomic library. We purified LipC12 with a N-terminal His-tag and then removed the tag using tobacco etch virus
Kamila Dmochowska et al.
Journal of insect science (Online), 13, 77-77 (2013-11-14)
Bees of the genus Osmia are being used in crop pollination at an increasing rate. However, a short life expectancy of adult individuals limits the feasibility of their use. Cocoons of the red mason bee, Osmia rufa L. (Hymenoptera: Megachilidae)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service