Skip to Content
Merck
All Photos(1)

Documents

M7920

Sigma-Aldrich

Minoxidil Sulfate

Synonym(s):

U-58838

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N5O4S
CAS Number:
Molecular Weight:
289.31
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.77

form

powder

originator

Johnson & Johnson

storage temp.

−20°C

SMILES string

Nc1cc(nc(N)[n+]1OS([O-])(=O)=O)N2CCCCC2

InChI

1S/C9H15N5O4S/c10-7-6-8(13-4-2-1-3-5-13)12-9(11)14(7)18-19(15,16)17/h6H,1-5H2,(H4,10,11,12,15,16,17)

InChI key

OEOLOEUAGSPDLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Minoxidil sulfate (MXS) has been used as a drug agent to study its effects on alopecia in corticotropin-releasing factor over-expressing (CRF-OE) mice. It has also been used as a positive control in an assay for the culturing of rat vibrissa follicles.

Biochem/physiol Actions

Minoxidil sulfate (MXS) is an endogenous derivative of minoxidil. It possesses greater aqueous solubility and is a potent vasodilator. MXS has the potential to treat androgenic alopecia or male baldness.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Active metabolite of minoxidil.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K D Meisheri et al.
The Journal of pharmacology and experimental therapeutics, 266(2), 655-665 (1993-08-01)
This study describes the in vitro and in vivo characteristics of a guanidine 4-morpholinecarboximidine-N-1-adamantyl-N'-cyclohexyl-hydroc hloride (U-37883A), as an antagonist of vascular ATP-sensitive K+ channels (KATP). In isolated rabbit mesenteric artery, the antagonistic effects of U-37883A (0.5-5 microM) were studied against
Michael J Shackcloth et al.
European journal of cardio-thoracic surgery : official journal of the European Association for Cardio-thoracic Surgery, 34(4), 839-844 (2008-08-06)
Vasodilator strategies used to treat bypass grafts in the operating theatre, such as nitrates, phosphodiesterase inhibitors and calcium channel antagonists have a broad but short-lived effect against a variety of vasoconstrictor stimuli. Treatments that react irreversibly with proteins modulating vasoconstriction
C E Ohrnberger et al.
The Journal of pharmacology and experimental therapeutics, 267(1), 25-30 (1993-10-01)
Glyburide, a sulfonylurea, and U-37883A, a guanidine (4-Morpholinecarboximidine-N-1-Adamantyl-N' cyclohexylhydrochloride), have been previously characterized as antagonists of the vascular ATP-sensitive K+ channels (KATP). In this report, the in vitro interaction between these two chemically distinct KATP antagonists was investigated using isolated
Pharmacological effects of drug conjugates: is morphine 6-glucuronide an exception?
G J Mulder
Trends in pharmacological sciences, 13(8), 302-304 (1992-08-01)
Nagendra S Ningaraj et al.
Cancer research, 63(24), 8899-8911 (2003-12-26)
Brain tumor microvessels/capillaries limit drug delivery to tumors by forming a blood-brain tumor barrier (BTB). The BTB overexpresses ATP-sensitive potassium (K(ATP)) channels that are barely detectable in normal brain capillaries, and which were targeted for BTB permeability modulation. In a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service