Skip to Content
Merck
All Photos(1)

Documents

Y0001432

Minoxidil for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Minoxidil, 6-(1-Piperidinyl)-2,4-pyrimidinediamine 3-oxide, 6-(1-Piperidinyl)pyrimidine-2,4-diamine 3-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N5O
CAS Number:
Molecular Weight:
209.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

minoxidil

manufacturer/tradename

EDQM

mp

272-274 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC1=CC(=NC(=N)N1O)N2CCCCC2

InChI

1S/C9H15N5O/c10-7-6-8(12-9(11)14(7)15)13-4-2-1-3-5-13/h6,11,15H,1-5,10H2

InChI key

ZIMGGGWCDYVHOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Minoxidil for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Activates ATP-activated K+ channels; vasodilator; slow or stop hair loss and promote hair regrowth.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Topical minoxidil for hair loss in women.
D Hong et al.
DICP : the annals of pharmacotherapy, 24(11), 1062-1063 (1990-11-01)
Marc R Avram et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 28(10), 894-900 (2002-11-02)
Over the last decade surgical management of hair loss has become an increasingly popular and satisfying procedure for both men and women, as innovations in donor harvesting, graft size, and hairline design have resulted in consistently natural-appearing hair restoration. In
T P Dooley
Experimental dermatology, 8(4), 328-329 (1999-08-10)
Cytosolic sulfotransferases (ST) catalyze the sulfation of various phenolic agents, catecholamines, thyroid hormones, steroids, drugs, and procarcinogens, usually resulting in the inactivation and subsequent excretion of the compound. My laboratory's efforts have focused on the cloning of the human phenol-sulfating
Susumu Otomo
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 119(3), 167-174 (2002-03-28)
The length and size of hair are depend on the anagen term in its hair cycle. It has been reported that the some cell growth factors, such as VEGF, FGF-5S, IGF-1 and KGF, induce the proliferation of cells in the
R L DeVillez
Dermatologic clinics, 8(2), 367-375 (1990-04-01)
Topical minoxidil is a trichogenic agent that stimulates the hair follicle via the vasoactive metabolite minoxidil sulfate without any evidence of antiandrogen activity or an effect on the immune system. Less than 5% of the applied dose is absorbed. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service