Skip to Content
Merck
All Photos(1)

Key Documents

M5693

Sigma-Aldrich

MEDICA 16

powder, ≥97% (HPLC)

Synonym(s):

3,3,14,14-Tetramethylhexadecanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H38O4
CAS Number:
Molecular Weight:
342.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to beige

mp

154-159 °C

solubility

DMSO: 10 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)(CCCCCCCCCCC(C)(C)CC(O)=O)CC(O)=O

InChI

1S/C20H38O4/c1-19(2,15-17(21)22)13-11-9-7-5-6-8-10-12-14-20(3,4)16-18(23)24/h5-16H2,1-4H3,(H,21,22)(H,23,24)

InChI key

HYSMCRNFENOHJH-UHFFFAOYSA-N

Biochem/physiol Actions

MEDICA 16 is an ATP-citrate lyase inhibitor and potent triacylglycerol-lowering agent. The product is hypolipidemic, antidiabetic, and shows reduced adiposity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Kalderon et al.
American journal of physiology. Endocrinology and metabolism, 279(1), E221-E227 (2000-07-14)
Adipose tissue lipolysis and fatty acid reesterification by liver and adipose tissue were investigated in rats fasted for 15 h under basal and calorigenic conditions. The fatty acid flux initiated by adipose fat lipolysis in the fasted rat is mostly
Qi Sun et al.
Molecular pharmacology, 78(5), 804-810 (2010-08-06)
GPR120 is a G protein-coupled receptor expressed preferentially in the intestinal tract and adipose tissue, that has been implicated in mediating free fatty acid-stimulated glucagon-like peptide-1 (GLP-1) secretion. To develop GPR120-specific agonists, a series of compounds (denoted as NCG compounds)
N Mayorek et al.
The Biochemical journal, 289 ( Pt 3), 911-917 (1993-02-01)
Treatment of cholesterol-fed male hamsters kept on a diet of purina chow with beta beta'-methyl-substituted hexadecanedioic acid (MEDICA 16) resulted in a progressive hypocholesterolaemic effect, amounting to a 50% decrease in the cholesterol content of all plasma lipoproteins. The decrease
J C Russell et al.
Diabetes, 47(5), 770-778 (1998-05-20)
The JCR:LA-cp rat develops an extreme obese/insulin-resistant syndrome such that by 12 weeks of age, there is no longer any insulin-mediated glucose turnover. At 4 weeks of age, obese and lean rats have essentially identical basal and insulin-mediated glucose uptake
O Hermesh et al.
The Journal of biological chemistry, 273(7), 3937-3942 (1998-03-28)
Mitochondria uncoupling by fatty acids in vivo is still questionable, being confounded by their dual role as substrates for oxidation and as putative genuine uncouplers of oxidative phosphorylation. To dissociate between substrate and the uncoupling activity of fatty acids in

Articles

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service