Skip to Content
Merck
All Photos(1)

Key Documents

M5280

Sigma-Aldrich

Mastoparan, Vespula lewisii

≥97% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H131N19O15
CAS Number:
Molecular Weight:
1478.91
Beilstein:
5491949
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](N)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)NC([C@@H](C)CC)C(=O)N[C@@H](CC(C)C)C(N)=O

InChI

1S/C70H131N19O15/c1-17-40(11)55(75)69(103)88-53(35-54(74)90)68(102)87-52(34-39(9)10)67(101)83-46(25-19-22-28-71)62(96)78-45(16)61(95)86-50(32-37(5)6)65(99)79-42(13)58(92)77-43(14)60(94)85-51(33-38(7)8)66(100)80-44(15)59(93)81-47(26-20-23-29-72)63(97)82-48(27-21-24-30-73)64(98)89-56(41(12)18-2)70(104)84-49(57(76)91)31-36(3)4/h36-53,55-56H,17-35,71-73,75H2,1-16H3,(H2,74,90)(H2,76,91)(H,77,92)(H,78,96)(H,79,99)(H,80,100)(H,81,93)(H,82,97)(H,83,101)(H,84,104)(H,85,94)(H,86,95)(H,87,102)(H,88,103)(H,89,98)/t40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,55-,56-/m0/s1

InChI key

MASXKPLGZRMBJF-MVSGICTGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Amino Acid Sequence

Ile-Asn-Leu-Lys-Ala-Leu-Ala-Ala-Leu-Ala-Lys-Lys-Ile-Leu-NH2

General description

Mastoparan is a cationic, amphiphilic, tetradecapeptide amide. It is composed of the restricted amino acid and the repeated sequence. This protein is a nonspecific secretagogue isolated from venom of wasp.

Application

Mastoparan has been used as a calmodulin antagonist.

Biochem/physiol Actions

Mast cell degranulating peptide. Activates phospholipase A2; inhibits calmodulin. Cell permeable; stimulates exocytosis and phosphoinositide breakdown.
Mastoparan effectively stimulates exocytosis from diverse mammalian cells. The encoded protein facilitates the secretion of histamine from mast cells, serotonin from platelets, catecholamines from chromaffin cells and prolactin from the anterior pituitary. In addition, it is also involved in activation of Phospholipase A2 and phosphoinositides (Pl) break down. Mastoparan functions as an inhibitor for calmodulin-stimulated phosphodiesterase. Mastoparan helps in mitochondrial permeability transition by enhancing an apparent bimodal mechanism of action.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new mast cell degranulating peptide "mastoparan" in the venom of Vespula lewisii.
Y Hirai et al.
Chemical & pharmaceutical bulletin, 27(8), 1942-1944 (1979-08-01)
K M Clapp et al.
American journal of physiology. Heart and circulatory physiology, 304(5), H660-H666 (2013-01-22)
Statin drugs inhibit 3-hydroxy-3-methylglutaryl CoA reductase, which reduces the synthesis of both cholesterol and isoprenoids (geranylgeranyl pyrophosphate and farnesyl pyrophosphate), with the latter being lipid molecules responsible for the posttranslational modification of small GTP-binding proteins such as Rho. Effects of
Sarah Jones et al.
Bioconjugate chemistry, 23(1), 47-56 (2011-12-14)
Retro-inverso transformation has commonly been employed as a strategy both for the synthesis of proteolytically stable peptide analogues and for the detailed investigation of structure activity relationships. Herein, we adopted a similar strategy to probe the structure activity relationships of
Asha Amritraj et al.
Endocrinology, 153(10), 4784-4798 (2012-08-21)
The IGF-II/mannose 6-phosphate (M6P) receptor is a single-transmembrane domain glycoprotein that plays an important role in the intracellular trafficking of lysosomal enzymes and endocytosis-mediated degradation of IGF-II. The receptor may also mediate certain biological effects in response to IGF-II binding
Activation of Phospholipase A2 by Auxin and Mastoparan in Hypocotyl Segments from Zucchini and Sunflower
Scherer GFE
Journal of Plant Physiology, 145, 483-490 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service