Skip to Content
Merck
All Photos(1)

Key Documents

IMPC-051-03

Supelco

Paraxanthine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

room temp

SMILES string

CN1C(=O)Nc2ncn(C)c2C1=O

InChI

1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)11(2)7(13)9-5/h3H,1-2H3,(H,9,13)

InChI key

QUNWUDVFRNGTCO-UHFFFAOYSA-N

General description

Paraxanthine is a major plasma and urinary metabolite of caffeine. This xanthine derivative is, along with caffeine, a central nervous system stimulant. This Snap-N-Spike® reference solution is applicable for caffeine quantitation in clinical toxicology or urine drug testing by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vidya Perera et al.
The Journal of pharmacy and pharmacology, 63(9), 1161-1168 (2011-08-11)
To investigate the utility of metrics of CYP1A2 activity using caffeine as a probe, and saliva and plasma sampling with or without a 24-h caffeine abstinence. This was a cross-over pharmacokinetic study in 30 healthy male subjects who received a
Rebeca Pinhancos et al.
Journal of mass spectrometry : JMS, 46(11), 1175-1181 (2011-11-30)
The presence of pharmaceuticals in drinking water is an emerging environmental concern. In most environmental testing laboratories, LC-MS/MS assays based on selected reaction monitoring are used as part of a battery of tests used to assure water quality. Although LC-MS/MS
Samanta Yubero-Lahoz et al.
Drug metabolism and pharmacokinetics, 27(6), 605-613 (2012-06-08)
3,4-Methylenedioxymethamphetamine (MDMA; ecstasy) is a ring-substituted amphetamine widely used for recreational purposes. MDMA is predominantly O-demethylenated in humans by cytochrome P450 (CYP) 2D6, and is also a potent mechanism-based inhibitor of the enzyme. After assessing the inhibition and recovery of
Saina Sharmin et al.
PloS one, 7(6), e38082-e38082 (2012-06-16)
Maternal caffeine consumption is associated with reduced fetal growth, but the underlying molecular mechanisms are unknown. Since there is evidence that decreased placental 11β-hydroxysteroid dehydrogenase type 2 (11β-HSD2) is linked to fetal growth restriction, we hypothesized that caffeine may inhibit
Vidya Perera et al.
Biomedical chromatography : BMC, 24(10), 1136-1144 (2010-09-21)
Caffeine has been extensively used as a probe to measure CYP1A2 activity in humans with caffeine clearance or the paraxanthine (major metabolite of caffeine) to caffeine concentration ratio being regarded as the preferred metric. A simple reverse-phased C(18) HPLC assay

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service