Skip to Content
Merck
All Photos(1)

Key Documents

M3199

Sigma-Aldrich

MK-212 hydrochloride

≥98% (HPLC)

Synonym(s):

2-Chloro-6-(1-piperazinyl)-pyrazine hydrochloride, 6-Chloro-2-(1-piperazinyl)pyrazine hydrochloride, NSC 31732

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11ClN4·HCl
CAS Number:
Molecular Weight:
235.11
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.Clc1cncc(n1)N2CCNCC2

InChI

1S/C8H11ClN4.ClH/c9-7-5-11-6-8(12-7)13-3-1-10-2-4-13;/h5-6,10H,1-4H2;1H

InChI key

PFIZGLUIYAZQFU-UHFFFAOYSA-N

Biochem/physiol Actions

Selective 5-HT2C serotonin receptor agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriane M dela Cruz et al.
Behavioural pharmacology, 20(8), 720-730 (2009-11-11)
Only a subgroup of human drug users progress from initial drug taking to drug addiction. The learned associations between the effects of the drug and the environment in which it is experienced is an important aspect of the progression to
M L Pierce et al.
Neuroscience, 170(3), 827-838 (2010-07-24)
Neuronal networks generating rhythmic activity as an emergent property are common throughout the nervous system. Some are responsible for rhythmic behaviours, as is the case for the spinal cord locomotor networks; however, for others the function is more subtle and
Martilias Farrell et al.
Psychopharmacology, 211(2), 149-159 (2010-06-08)
The extended ternary complex theory of receptor function states a ligand can be classified as an inverse agonist, agonist, or neutral antagonist based on its ability to preferentially stabilize the inactive conformation, the active conformation, or to have no preference
N K Popova et al.
Neuroscience and behavioral physiology, 40(4), 357-361 (2010-03-26)
The functional activity of serotonin 5-HT(2A) and 5-HT(2C) receptors and the expression of the genes encoding them were studied in Norway rats bred for 60 generations for the presence and absence of high levels of stress-evoked aggression to humans. There
Peter Salchner et al.
Psychopharmacology, 185(3), 282-288 (2006-03-08)
We have recently reported that acute treatment with the selective serotonin reuptake inhibitor fluoxetine exacerbates escape responses to airjet and facilitates airjet-induced activation of locus coeruleus (LC) neurons. Here we aimed to identify the 5-HT receptor subtype(s) mediating the anxiogenic-like

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service