Skip to Content
Merck
All Photos(1)

Key Documents

A8727

Sigma-Aldrich

Antalarmin hydrochloride

≥98% (HPLC), solid, CRF1 antagonist

Synonym(s):

N-Butyl-N-ethyl-2,5,6-trimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolol[2,3-d]pyrimidin-4-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34N4·HCl
CAS Number:
Molecular Weight:
415.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Antalarmin hydrochloride, ≥98% (HPLC), solid

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].CCCCN(CC)c1nc(C)nc2n(c(C)c(C)c12)-c3c(C)cc(C)cc3C

InChI

1S/C24H34N4.ClH/c1-9-11-12-27(10-2)23-21-18(6)19(7)28(24(21)26-20(8)25-23)22-16(4)13-15(3)14-17(22)5;/h13-14H,9-12H2,1-8H3;1H

InChI key

CGDGXEDXEXACKQ-UHFFFAOYSA-N

Application

Antalarmin hydrochloride has been used:
  • to study its effect on stress-induced tryptophan hydroxylase 2 activity
  • to study the anxiogenic effects of corticotropin-releasing factor 1 (CRF1) activation
  • to analyze the therapeutic effect of blocking corticotrophin-releasing hormone (CRH) signaling in treating social stress-induced depression

Antalarmin hydrochloride is a non-peptide CRF1 corticotropin-releasing factor receptor antagonist.

Biochem/physiol Actions

Antalarmin serves as a non steroidal inhibitor of pregnancy even during the early stages. It is known to delay parturition. It also prevents the development of gastric ulcers which arises in response to stress.
Antalarmin hydrochloride is a non-peptide CRF1 corticotropin-releasing factor receptor antagonist.

Features and Benefits

This compound is featured on the Corticotropin-Releasing Factor Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pediatric Neuroendocrinology, 41-41 (2010)
Cecile Ducottet et al.
Progress in neuro-psychopharmacology & biological psychiatry, 27(4), 625-631 (2003-06-06)
Several recent studies on corticotropin-releasing factor (CRF) have suggested that this neuropeptide may play a role in depression. Consequently, CRF receptor antagonists have been proposed as potential new agents for the treatment of this condition. This study investigated the effects
CRF type 1 receptors of the medial amygdala modulate inhibitory avoidance responses in the elevated T-maze
Vicentini JE, et al.
Hormones and Behavior, 65(3), 195-202 (2014)
Deficits in KCC2 and activation of the HPA axis lead to depression-like behavior following social defeat
Miller S and Maguire J
Hormonal Studies, 2(1), 2-2 (2014)
Rodney C Samaco et al.
Nature genetics, 44(2), 206-211 (2012-01-11)
Genomic duplications spanning Xq28 are associated with a spectrum of phenotypes, including anxiety and autism. The minimal region shared among affected individuals includes MECP2 and IRAK1, although it is unclear which gene when overexpressed causes anxiety and social behavior deficits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service