Skip to Content
Merck
All Photos(2)

Key Documents

M0508

Sigma-Aldrich

Micrococcus lysodeikticus ATCC No. 4698

lyophilized cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.51

form

lyophilized cells

Quality Level

storage temp.

−20°C

Other Notes

A source of DNA.

Preparation Note

Washed in 0.01 M EDTA, pH 7.0, and 0.15 M sodium chloride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Buist et al.
Journal of bacteriology, 177(6), 1554-1563 (1995-03-01)
A gene of Lactococcus lactis subsp. cremoris MG1363 encoding a peptidoglycan hydrolase was identified in a genomic library of the strain in pUC19 by screening Escherichia coli transformants for cell wall lysis activity on a medium containing autoclaved, lyophilized Micrococcus
C Potvin et al.
Molecular & general genetics : MGG, 214(2), 241-248 (1988-10-01)
Several hundred bacterial isolates were screened for bacteriolytic activity by growing them on agar medium containing autoclaved, lyophilized Micrococcus lysodeikticus cells as the substrate. A Bacillus sp. producing the largest lytic zone was selected. A genomic bank of this selected
Soichiro Nakamura et al.
Bioconjugate chemistry, 17(5), 1170-1177 (2006-09-21)
Bioactive oligomannosyl lysozyme with improved surface functionalities was successfully prepared by using an extracellular pH-sensitive glycosylation system for heterogeneous protein in yeast cell. A recombinant Saccharomyces cerevisiae carrying a mutant lysozyme gene encoding the signal sequence of an N-linked glycosylation
Covalently closed circular DNA from Micrococcus lysodeikticus cells infected with phage N1.
C S Lee et al.
Biochemical and biophysical research communications, 32(5), 752-756 (1968-09-06)
Fatma M Abdelhamid et al.
Environmental science and pollution research international, 27(10), 10950-10965 (2020-01-19)
Lead, toxic heavy metal of global concern, induces toxicity in various organs via oxidative stress. Thereby, in this study, the protective role of curcumin against lead acetate-induced toxicity was evaluated. Thirty-two male albino rats were allocated equally into four groups

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service