Skip to Content
Merck
All Photos(1)

Key Documents

10837059001

Roche

Lysozyme

from hen egg white

Synonym(s):

Mucopeptide N-acetylmuramoyl-hydrolase, Muramidase, lysozyme

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

chicken egg white

Quality Level

Assay

≥95%

form

solid

specific activity

>23000 U/mg (Shugar units)

mol wt

14.4 kDa

packaging

pkg of 10 g

manufacturer/tradename

Roche

optimum pH

6.0-7.0

absorption

3.9 at 280 nm (10 mg enzyme/ml)

Related Categories

General description

Muramidase, Mucopeptide N-acetylmuramoyl-hydrolase

Application

Lysozyme has been used for:
  • Cell wall degradation
  • Preparation of protoplasts
  • Bacteriolysis
  • Pharmacology (anti-inflammatory effect on mucous membrane; healing of tissue; protection against bleeding; normalization of mucus secretion; dissolution of pyrogens; potentiation of antibiotics)
  • Food and drinks (flavor enhancer)
  • Sample preparation prior to isolation of nucleic acids
Lysozyme has been used in the monitoring of transcription by real-time -PCR and sample preparation.

Biochem/physiol Actions

Lysozyme hydrolytically cleaves the bonds between N-acetyl-D-glucosamine and N-acetylmuramic acid residues (GlcNAc β1-4Mur) in mucopolysaccharides and the glycan skeletons of the corresponding peptidoglycans (the latter are glycosaminoglycans linked to or crosslinked by peptide chains). Lysozyme effectively destroys the cell walls of bacteria.

Unit Definition

One Shugar unit is that quantity of enzyme in 1 ml of a suspension of Micrococcus luteus of pH 7.0 whose initial absorbance at a wavelength of 450 nm is 0.750 for a pathlength of 10 mm which causes the absorbance to decrease at the rate of 0.001 per minute, all measurements being carried out at a temperature of +25 °C.

Preparation Note

Working solution: Lysozyme hydrochloride is readily soluble in water and buffer solutions but insoluble in organic solvents.
Storage conditions (working solution): -15 to -25 °C
Aqueous solutions of the hydrochloride (2 mg/ml dist. water) can be stored for several days at 2 to 8° C or at -15 to -25° C for several weeks.

Reconstitution

Prepare a stock solution at a concentration of 50 mg/ml in dist. water. Dispense into aliquots and store at -15 to -25 °C. Discard each aliquot after use.

Storage and Stability

Store at 2 to 8 °C. (Store dry!)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning and expression of metagenomic DNA in Streptomyces lividans and subsequent fermentation for optimized production
Yuriy R, et al.
Meta Gene (2017)
Lorena Hentschel et al.
Physical chemistry chemical physics : PCCP, 23(4), 2686-2696 (2021-01-23)
During a first-order phase transition, a thermodynamic system releases or absorbs latent heat. Despite their fundamental importance, the heat or enthalpy change occurring during protein crystallization has been directly measured only in a few cases, and the associated entropy change
Blanca López-Méndez et al.
European biophysics journal : EBJ, 50(3-4), 411-427 (2021-04-22)
Microscale thermophoresis (MST), and the closely related Temperature Related Intensity Change (TRIC), are synonyms for a recently developed measurement technique in the field of biophysics to quantify biomolecular interactions, using the (capillary-based) NanoTemper Monolith and (multiwell plate-based) Dianthus instruments. Although
Isolation of Heterocysts from Anabaena sp. PCC 7120
Maria Ermakova* and Yagut Allahverdiyeva
Bio-protocol, 5(8) (2015)
A new iron acquisition system in Bacteroidetes.
Manfredi P, et al.
Infection and Immunity (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service