Skip to Content
Merck
All Photos(1)

Key Documents

LO4200

Sigma-Aldrich

LOPAC®1280 - Small Scale

International Version

Synonym(s):

Library of Pharmacologically Active Compounds - small scale

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

form

liquid

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Compounds are arranged in 96-well format in 16 racks of 80, one compound per well (25 μl at 10 mM in DMSO in archival storage racks).

Application

Library of pharmacologically active compounds LOPAC®1280 - small scale has been used to screen for the suppressors of niclosamide- and nigericin-induced interleukin (IL-1β) release.

Features and Benefits

Includes the latest, drug-like molecules in the fields of Cell Signaling & Neuroscience:

  • Apoptosis
  • G Proteins & Cyclic Nucleotides
  • Gene Regulation & Expression
  • Ion Channels
  • Lipid Signaling
  • Multi-Drug Resistance
  • Neurotransmission
  • Phosphorylation
SDFile provided to construct a database containing:
  • Structure
  • Primary Name
  • Secondary Name
  • Pharmacological Activity
  • Product Number
  • Rack Position

Other Notes

For sale outside of the U.S. only. U.S. customers should refer to Product Code LO4100.

Legal Information

LOPAC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Lact. - Muta. 1B - Repr. 1A - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Laura M Alcântara et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-04)
High genetic and phenotypic variability between Leishmania species and strains within species make the development of broad-spectrum antileishmanial drugs challenging. Thus, screening panels consisting of several diverse Leishmania species can be useful in enabling compound prioritization based on their spectrum
Anzar Asad et al.
Basic & clinical pharmacology & toxicology, 133(4), 364-377 (2023-07-03)
Adhesion G protein-coupled receptors (GPCRs) are an underrepresented class of GPCRs in drug discovery. We previously developed an in vivo drug screening pipeline to identify compounds with agonist activity for Adgrg6 (Gpr126), an adhesion GPCR required for myelination of the
Niclosamide activates the NLRP3 inflammasome by intracellular acidification and mitochondrial inhibition
Tran UT and Kitami T
Communications biology, 2(1), 2-2 (2019)
High-throughput screening identifies compounds that enhance lentiviral transduction.
Johnston JM, Denning G, Moot R, et al.
Gene Therapy, doi:10-doi:10 (2014)
Nicolas Wyhs et al.
Journal of biomolecular screening, 19(7), 1060-1069 (2014-03-13)
Methylated DNA binding proteins such as Methyl-CpG Binding Domain Protein 2 (MBD2) can transduce DNA methylation alterations into a repressive signal by recruiting transcriptional co-repressor complexes. Interfering with MBD2 could lead to reactivation of tumor suppressor genes and therefore represents

Articles

Explore compound library screening options with our Pharmacologically Active Compounds portfolio.

Explore compound library screening options with our Pharmacologically Active Compounds portfolio.

Explore compound library screening options with our Pharmacologically Active Compounds portfolio.

Explore compound library screening options with our Pharmacologically Active Compounds portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service