Skip to Content
Merck
All Photos(1)

Key Documents

L7134

Sigma-Aldrich

Luteinizing hormone releasing hormone human acetate salt

≥98% (HPLC), powder

Synonym(s):

Gonadoliberin, Gonadotropin releasing hormone, LH-RH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H75N17O13
CAS Number:
Molecular Weight:
1182.29
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

powder

mol wt

Mr 1182.33

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(O)=O.CC(O)=O.CC(C)C[C@H](NC(=O)CNC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](CO)NC(=O)[C@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4c[nH]cn4)NC(=O)[C@@H]5CCC(=O)N5)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N6CCC[C@H]6C(=O)NCC(N)=O

InChI

1S/C55H75N17O13.2C2H4O2/c1-29(2)19-38(49(80)67-37(9-5-17-60-55(57)58)54(85)72-18-6-10-43(72)53(84)62-25-44(56)75)66-46(77)26-63-47(78)39(20-30-11-13-33(74)14-12-30)68-52(83)42(27-73)71-50(81)40(21-31-23-61-35-8-4-3-7-34(31)35)69-51(82)41(22-32-24-59-28-64-32)70-48(79)36-15-16-45(76)65-36;2*1-2(3)4/h3-4,7-8,11-14,23-24,28-29,36-43,61,73-74H,5-6,9-10,15-22,25-27H2,1-2H3,(H2,56,75)(H,59,64)(H,62,84)(H,63,78)(H,65,76)(H,66,77)(H,67,80)(H,68,83)(H,69,82)(H,70,79)(H,71,81)(H4,57,58,60);2*1H3,(H,3,4)/t36-,37-,38-,39-,40-,41-,42-,43-;;/m0../s1

InChI key

QOVPLTBEZAJSHY-VIRDRFNDSA-N

Gene Information

human ... GNRH1(2796)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2

Application

Luteinizing hormone releasing hormone human acetate salt has been used:
  • for synchronization of the ovulations
  • for pre-absorption of the anti-GnRH antibody in staining procedures
  • to study pituitary sensitivity to GnRH in presence of endocrine disrupting compound

Biochem/physiol Actions

Hypothalamic peptide that stimulates release of gonadotrophins from anterior pituitary, thus regulating reproductive functions.
Stimulates release of gonadotrophins from anterior pituitary.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Components

pGlu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2

Other Notes

free peptide
Lyophilized from 0.5% acetic acid

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Developmental Programming: Impact of Prenatal Exposure to Bisphenol-A and Methoxychlor on Steroid Feedbacks in Sheep
Bachir Abi Salloum
Toxicology and Applied Pharmacology (2013)
Nicholas A Levis et al.
Ecology and evolution, 5(7), 1405-1411 (2015-04-22)
Phenotypic plasticity is commonplace, and plasticity theory predicts that organisms should often evolve mechanisms to detect and respond to environmental cues that accurately predict future environmental conditions. Here, we test this prediction in tadpoles of spadefoot toads, Spea multiplicata. These
Verónica Berta Dorfman et al.
Biology of reproduction, 89(5), 115-115 (2013-10-04)
In mammals, elevated levels of progesterone (P4) throughout gestation maintain a negative feedback over the hypothalamic-hypophyseal-gonadal (H-H-G) axis, avoiding preovulatory follicular growth and preventing ovulation. Recent studies showed that in the South American plains vizcacha (Lagostomus maximus) folliculogenesis progresses to
Histological characterization of gonadotropin-releasing hormone
(GnRH) in the hypothalamus of the South American plains
vizcacha (Lagostomus maximus)
Veronica, et.la
Journal of Molecular Histology (2011)
First birth of an animal from an extinct subspecies
(Capra pyrenaica pyrenaica) by cloning
J. Folch
Theriogenology (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service