Skip to Content
Merck
All Photos(1)

Key Documents

E3888

Sigma-Aldrich

[D-Pen2,5]-Enkephalin hydrate

≥95% (HPLC)

Synonym(s):

DPDPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H39N5O7S2 · xH2O
Molecular Weight:
645.79 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

CC1(C)SSC(C)(C)[C@@H](NC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)NCC(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@H]1C(O)=O

InChI

1S/C30H39N5O7S2/c1-29(2)23(34-25(38)20(31)14-18-10-12-19(36)13-11-18)27(40)32-16-22(37)33-21(15-17-8-6-5-7-9-17)26(39)35-24(28(41)42)30(3,4)44-43-29/h5-13,20-21,23-24,36H,14-16,31H2,1-4H3,(H,32,40)(H,33,37)(H,34,38)(H,35,39)(H,41,42)/t20-,21-,23-,24-/m0/s1

InChI key

MCMMCRYPQBNCPH-WMIMKTLMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Amino Acid Sequence

Tyr-Pen-Gly-Phe-Pen [Disulfide Bridge: 2-5]

General description

[D-Pen2,5]-Enkephalin hydrate, also called DPDPE, is a peptide and acts as a δ1-opioid receptor agonist. Opioid receptors are divided into three types called, μ, κ and δ, depending upon their ligands. These ligands are peptides and are classified as enkephalins, endorphins and dynorphins.

Application

[D-Pen2,5]-Enkephalin hydrate (DPDPE) has been used to study the pharmacodynamic and pharmacokinetic capabilities of polyethylene glycol (PEG) conjugate of DPDPE in rodents. It has been used to study the tendency of opiate self-administration in male Long-Evans rats, in ventral tegmental area (VTA).

Biochem/physiol Actions

Antinociceptive activity mediated through the δ1 receptor while the modulatory activity is mediated through the δ2 receptor. Tritiated [D-Pen2,5]-enkephalin is used as a δ1 ligand.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D C Jewett et al.
Psychopharmacology, 127(3), 225-230 (1996-10-01)
The present study assessed the discriminative stimulus effects of the delta-opioid agonist [D-Pen2-D-Pen5]enkephalin (DPDPE) in pigeons. Food-restricted pigeons were trained to discriminate between i.c.v injections of 100 micrograms [D-Pen2-D-Pen5]enkephalin (DPDPE) and saline in a two-key operant procedure; acquisition of discriminative
S Allouche et al.
Biochemical pharmacology, 59(8), 915-925 (2000-02-29)
The two pharmacological delta-opioid receptor subtypes, delta1 and delta2, have been defined on the basis of pharmacological tools but remain to be characterized at the molecular level, since only a single cDNA has been cloned. The present study aimed to
Maria Camilla Cerlesi et al.
European journal of pharmacology, 794, 115-126 (2016-11-23)
An innovative chemical strategy named peptide welding technology (PWT) has been developed for the facile synthesis of tetrabranched peptides. [Dmt1]N/OFQ(1-13)-NH2 acts as a universal agonist for nociceptin/orphanin FQ (N/OFQ) and classical opioid receptors. The present study investigated the pharmacological profile
Dinah Nockemann et al.
EMBO molecular medicine, 5(8), 1263-1277 (2013-07-03)
The use of opioid agonists acting outside the central nervous system (CNS) is a promising therapeutic strategy for pain control that avoids deleterious central side effects such as apnea and addiction. In human clinical trials and rat models of inflammatory
K Akiyama et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(8), 2543-2547 (1985-04-01)
Specific binding properties of the tritium-labeled delta opiate receptor agonist [3H][2-D-penicillamine, 5-D-penicillamine]enkephalin [( 3H][D-Pen2, D-Pen5]enkephalin) were characterized in the rat brain and in a mouse neuroblastoma-rat glioma hybrid cell line (NG 108-15). Saturation isotherms of [3H][D-Pen2, D-Pen5]enkephalin binding to rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service