Skip to Content
Merck
All Photos(2)

Key Documents

L6503

Sigma-Aldrich

Methyl γ-linolenate

≥99% (GC)

Synonym(s):

γ-Linolenic acid methyl ester, ω-6 Linolenic acid methyl ester, 6,9,12-Octadecatrienoic acid methyl ester, Fame 18:3n-6, Methyl (Z,Z,Z)-6,9,12-octadecatrienoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)3(CH2)4CO2CH3
CAS Number:
Molecular Weight:
292.46
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant seeds (Borage)

Assay

≥99% (GC)

form

liquid

refractive index

n20/D 1.476 (lit.)

bp

162 °C/0.5 mmHg (lit.)

functional group

ester

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/CCCCC(=O)OC

InChI

1S/C19H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h7-8,10-11,13-14H,3-6,9,12,15-18H2,1-2H3/b8-7-,11-10-,14-13-

InChI key

JFRWATCOFCPIBM-JPFHKJGASA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Isolation of methyl gamma linolenate from Spirulina platensis using flash chromatography and its apoptosis inducing effect.: This study explores the extraction and potential anticancer effects of methyl gamma-linolenate from Spirulina platensis by using it as a standard for identification, suggesting its use in developing natural anticancer compounds (Jubie et al., 2015).

Biochem/physiol Actions

Methyl γ-linolenate (C18:6,9,12) differs from methyl α-linolenate (C18:9,12,15) in the positions of the double bonds. Methyl γ-linolenate may be used in nutritional studies regarding weight regain and as a possible tumor suppression agent.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie A Schirmer et al.
The Journal of nutrition, 137(6), 1430-1435 (2007-05-22)
The purpose of this study was to determine whether gamma-linolenate (GLA) supplementation would suppress weight regain following major weight loss. Fifty formerly obese humans were randomized into a double-blind study and given either 890 mg/d of GLA (5 g/d borage
Michael M Meguid et al.
Nutrition (Burbank, Los Angeles County, Calif.), 24(9), 832-842 (2008-08-30)
Roux-en-Y gastric bypass (RYGB) produces rapid and dramatic weight loss in very heavy obese patients. Up to 20% cannot sustain their weight loss beyond 2 to 3 y after surgery. To identify putative etiologic factors producing post-RYGB weight regain, a
S Jubie et al.
BMC complementary and alternative medicine, 15, 263-263 (2015-08-05)
Isolation of methyl gamma linolenate from Spirulina platensis using flash chromatography and its apoptosis inducing effect against human lung carcinoma A- 549 cell lines. Gamma linolenic acid is an important omega-6 polyunsaturated fatty acid (PUFA) of medicinal interest was isolated
Hung Pham et al.
Prostaglandins, leukotrienes, and essential fatty acids, 74(4), 271-282 (2006-03-29)
Prostate cancer poses considerable threat to the aging male population as it has become a leading cause of cancer death to this group. Due to the complexity of this age-related disease, the mechanism(s) and factors resulting in prostate cancer remain
Jim-Wen Liu et al.
Journal of agricultural and food chemistry, 52(12), 3960-3966 (2004-06-10)
Dietary supplementation of a high-gamma-linolenic acid canola oil (HGCO) containing approximately 36% (w/w) of gamma-linolenic acid (GLA, 18:3n-6) from the seeds of a genetically transformed canola strain, was assessed for its long-term biological effects. Growing Sprague-Dawley rats (n = 30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service