Skip to Content
Merck
All Photos(2)

Documents

E4762

Sigma-Aldrich

Methyl elaidate

≥99% (capillary GC)

Synonym(s):

Elaidic acid methyl ester, Methyl trans-9-octadecenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOCH3
CAS Number:
Molecular Weight:
296.49
Beilstein:
1727038
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (capillary GC)

form

liquid

density

0.871 g/mL at 20 °C (lit.)

functional group

ester

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10+

InChI key

QYDYPVFESGNLHU-ZHACJKMWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byung-Jun Kollbe Ahn et al.
Journal of agricultural and food chemistry, 60(9), 2179-2189 (2012-02-07)
Toxic solvent and strong acid catalysts causing environmental issues have been mainly used for ring-opening of epoxidized oleochemicals. Here, we demonstrated that magnesium stearate (Mg-stearate) was a high efficient catalyst for solvent-free ring-opening of epoxidized methyl oleate, a model compound
Cyril Thurier et al.
ChemSusChem, 1(1-2), 118-122 (2008-07-09)
Unsaturated vegetable oils are an attractive renewable feedstock, and the selective cleavage of unsaturated fatty esters is an important transformation in this respect. The efficient and selective cross-metathesis of methyl oleate with ethylene was achieved under mild conditions with ruthenium-alkylidene
Ursula Biermann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(26), 8201-8207 (2012-05-18)
A kinetic study of the dodecanethiol-catalyzed cis/trans isomerization of methyl oleate (cis-2) without added initiator was performed by focusing on the initiation of the radical chain reaction. The reaction orders of the rate of isomerization were 2 and 0.5 for
Chinmay A Deshmane et al.
Chemical communications (Cambridge, England), 46(34), 6347-6349 (2010-08-18)
Thermally stable mesoporous Ga-Nb mixed oxides, active in both acid-catalysed and redox reactions have been synthesized via self-assembly hydrothermal assisted approach. Methyl oleate, a major component of biodiesels, undergoes double bond and skeletal isomerisation as well as dehydrogenation over these
Renee M Thomas et al.
Journal of the American Chemical Society, 133(19), 7490-7496 (2011-04-23)
N-Aryl,N-alkyl N-heterocyclic carbene (NHC) ruthenium metathesis catalysts are highly selective toward the ethenolysis of methyl oleate, giving selectivity as high as 95% for the kinetic ethenolysis products over the thermodynamic self-metathesis products. The examples described herein represent some of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service