Skip to Content
Merck
All Photos(3)

Key Documents

L5647

Sigma-Aldrich

Lidocaine hydrochloride monohydrate

≥99% (HPLC), solid, Na⁺ channel blocker

Synonym(s):

2-Diethylamino-N-(2,6-dimethylphenyl)acetamide hydrochloride monohydrate, Lignocaine hydrochloride monohydrate, Xylocaine hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O · HCl · H2O
CAS Number:
Molecular Weight:
288.81
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Lidocaine hydrochloride monohydrate, solid

form

solid

color

white

solubility

H2O: soluble

SMILES string

Cl[H].[H]O[H].CCN(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C14H22N2O.ClH.H2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4;;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H;1H2

InChI key

YECIFGHRMFEPJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lidocaine hydrochloride monohydrate is a commonly used as local anesthetic in topical applications and oral mucosal biopsies. It has a short anesthetic half-life.

Application

Lidocaine hydrochloride monohydrate has been used:
  • in the preparation of drug composite for the controlled delivery in vitro, evaluated by drug release profile in ultra-high performance liquid chromatography (UPLC) and as a reference standard for fourier transform infrared spectroscopy (FT-IR).
  • to record dose dependent response of sodium voltage (Nav1.7) channels in the injection-molded polymer device.
  • For inhibition of glial cells in brain tissue samples.

Biochem/physiol Actions

Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration
Na+ channel blocker; class IB antiarrhythmic.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis R Sparta et al.
Nature protocols, 7(1), 12-23 (2011-12-14)
In vivo optogenetic strategies have redefined our ability to assay how neural circuits govern behavior. Although acutely implanted optical fibers have previously been used in such studies, long-term control over neuronal activity has been largely unachievable. Here we describe a
Kristine von Krogh et al.
Biology, 10(11) (2021-11-28)
Zebrafish are often euthanized by overdose of anaesthesia. However, fish may have aversion towards some anaesthetics, and protocol efficacy varies between species. Using wild type adult Danio rerio, we assessed time to loss of opercular beat, righting, and startle reflexes
K Yokawa et al.
Annals of botany, 122(5), 747-756 (2017-12-14)
Anaesthesia for medical purposes was introduced in the 19th century. However, the physiological mode of anaesthetic drug actions on the nervous system remains unclear. One of the remaining questions is how these different compounds, with no structural similarities and even
Functionalized silver doped hydroxyapatite scaffolds for controlled simultaneous silver ion and drug delivery
Dubnika A, et al.
Ceramics International, 43(4), 3698-3705 (2017)
Julie I R Labau et al.
Frontiers in pharmacology, 12, 791740-791740 (2022-01-08)
Lacosamide, developed as an anti-epileptic drug, has been used for the treatment of pain. Unlike typical anticonvulsants and local anesthetics which enhance fast-inactivation and bind within the pore of sodium channels, lacosamide enhances slow-inactivation of these channels, suggesting different binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service