Skip to Content
Merck
All Photos(5)

Key Documents

L2250

Sigma-Aldrich

Lithium L-lactate

≥98% (titration)

Synonym(s):

(S)-2-Hydroxypropionic acid lithium salt, L-Lactic acid lithium salt, Sarcolactic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5LiO3
CAS Number:
Molecular Weight:
96.01
Beilstein:
6127244
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (titration)

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

<2% water (Karl Fischer)

color

white

solubility

water: 100 mg/mL, clear, colorless

cation traces

Li: 5.26-9.30% (anhydrous)

application(s)

food and beverages
hematology
microbiology

storage temp.

2-8°C

SMILES string

[Li+].C[C@H](O)C([O-])=O

InChI

1S/C3H6O3.Li/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1

InChI key

GKQWYZBANWAFMQ-DKWTVANSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium L-lactate, also known as Sarcolactic acid lithium or Lithium (S)-2-hydroxypropanoate, is a lithium salt of L-lactic acid, the optically active isomer of lactic acid characterized by an (S) stereochemical arrangement. L-Lactic acid is naturally produced in animals through the fermentation of pyruvate via lactate dehydrogenase during normal metabolism and exercise. This compound is a crucial element in lactic acid fermentation carried out by lactic acid bacteria, where carbohydrates are converted into lactic acid. In humans, lactic acid and its conjugate base play vital roles in various biological processes, spanning from prenatal development to the maintenance of the extracellular fluid surrounding neurons.

Recent research indicates that Lithium L-lactate exhibits mood-stabilizing effects, particularly in individuals with bipolar disorder. This impact is believed to be linked to its influence on the phosphatidylinositol (PI) pathway and the inhibition of glycogen synthase kinase-3 (GSK-3) activity. Beyond bipolar disorder, ongoing investigations are exploring the potential applications of Lithium L-lactate in neuropsychiatric conditions, including studies related to depression, anxiety, and schizophrenia, highlighting its significance in the areas of metabolomics and biochemical research.

Application

Lithium L-lactate has been used as a standard:
  • for the quantification of lactic acid using analytical methods
  • for the quantification of lactic acid using high-performance liquid chromatography
  • as a standard for L-lactic acid assay

Lithium L-lactate is a versatile compound that finds application in metabolomics and biochemical research.

Features and Benefits

  • Ideal for Metabolomics, Biochemical and Cell Biology research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Lebouvier et al.
Marine drugs, 7(4), 640-653 (2010-01-26)
As part of our search for new antimalarial drugs in South Pacific marine sponges, we have looked for inhibitors of Pfnek-1, a specific protein kinase of Plasmodium falciparum. On the basis of promising activity in a preliminary screening, the ethanolic
G Díaz-Ruiz et al.
Applied and environmental microbiology, 69(8), 4367-4374 (2003-08-07)
Pozol is an acid beverage obtained from the natural fermentation of nixtamal (heat- and alkali-treated maize) dough. The concentration of mono- and disaccharides from maize is reduced during nixtamalization, so that starch is the main carbohydrate available for lactic acid
Chia-Yu Lin et al.
PloS one, 15(10), e0241053-e0241053 (2020-10-27)
The relationship between methylglyoxal (MGO) and D-lactate during saikosaponin C (SSC) treatment of mice with accelerated nephrotoxic serum (NTS) nephritis was investigated. NTS nephritis was induced by administration of anti-basement membrane antibodies to C57BL/6 mice and three dosages of SSC
Mathias Busch et al.
Environmental research, 193, 110536-110536 (2020-12-01)
The continuous degradation of plastic waste in the environment leads to the generation of micro- and nanoplastic fragments and particles. Due to the ubiquitous presence of plastic particles in natural habitats as well as in food, beverages and tap water
Akram Abouie Mehrizi et al.
Medical microbiology and immunology, 207(2), 151-166 (2018-02-06)
Malaria vaccine development has been confronted with various challenges such as poor immunogenicity of malaria vaccine candidate antigens, which is considered as the main challenge. However, this problem can be managed using appropriate formulations of antigens and adjuvants. Poly(I:C) is

Protocols

Quick and accurate analysis of D- and L-lactic acid in dairy products using Reflectoquant® Lactic Acid Test upon reaction with lactate dehydrogenase (LDH).

Quick and accurate analysis of D- and L-lactic acid in dairy products using Reflectoquant® Lactic Acid Test upon reaction with lactate dehydrogenase (LDH).

Rapid analysis of DL-lactic acid in vegan yogurt using the Reflectoquant® Lactic Acid Test, and enzymatic reaction with Lactate Dehydrogenase (LDH).

Quick and accurate analysis of D- and L-lactic acid in dairy products using Reflectoquant® Lactic Acid Test upon reaction with lactate dehydrogenase (LDH).

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service