Skip to Content
Merck
All Photos(3)

Key Documents

KCQS00

Sigma-Aldrich

KiCqStart® SYBR® Green qPCR ReadyMix

For Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems

Synonym(s):

qPCR master mix, sybr green qPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

form

liquid

usage

sufficient for 1250 reactions
sufficient for 250 reactions
sufficient for 5000 reactions

feature

dNTPs included
hotstart

storage condition

protect from light

technique(s)

qPCR: suitable

color

colorless

input

purified DNA

compatibility

for use with Bio-Rad CFX384
for use with Bio-Rad CFX96
for use with Bio-Rad MJ Chromo4
for use with Bio-Rad MJ Opticon 2
for use with Bio-Rad MJ Opticon Cepheid SmartCycler
for use with Bio-Rad MiniOpticon
for use with Bio-Rad MyiQ
for use with Eppendorf® Mastercycler ep realplex2 s
for use with Eppendorf® Mastercycler ep realplex
for use with Illumina Eco qPCR
for use with Qiagen Corbett Rotor-Gene 3000
for use with Qiagen Corbett Rotor-Gene 6000
for use with Qiagen Corbett Rotor-Gene Q
for use with Roche LightCycler 480

detection method

SYBR® Green

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KiCqStart SYBR Green qPCR ReadyMix is a 2X concentrated, ready-to-use master mix that contains all components, except primers and template, for real-time quantitative PCR (qPCR) This unique combination of proprietary buffer, stabilizers, and Hot-Start Taq DNA polymerase delivers maximum PCR efficiency, sensitivity, specificity and robust fluorescent signal using fast, or conventional, cycling protocols with SYBR Green qPCR.

Highly specific amplification is crucial to successful qPCR with SYBR Green I dye technology because this dye binds to and detects any dsDNA generated during amplification. Hot-Start Taq DNA polymerase is antibody mediated to be inactive prior to the initial PCR denaturation step.

Application

KiCqStart® SYBR® Green qPCR ReadyMix has been used to perform quantitative real-time PCR (qPCR) for the amplification and quantification of genomic DNA extracted from human gut bacterium Prevotella copri.
PCR applications:
  • Gene expression
  • DNA quantification
  • CHiP

Features and Benefits

  • Assay results in as little as 33 minutes
  • Highly efficient and sensitive real-time PCR results
  • Little/no optimization required

Components

2X reaction buffer containing optimized concentrations of MgCl2, dNTPs (dATP, dCTP, dGTP, dTTP), KiCqStart Taq DNA Polymerase, SYBR Green dye, and stabilizers

packaging:
250 reactions* = 2 X 1.25 mL tubes
1250 reactions* = 10 X 1.25 mL tubes
5000 reactions* = 1 X 50 mL tube
*number of reactions based on a 20uL volume

Other Notes

Storage Conditions:
KiCqStart SYBR Green qPCR ReadyMix is stable for 1 year when stored in a constant temperature freezer at -20°C, protected from light. For convenience, it may be stored unfrozen at +2 to +8°C for up to 6 months. After thawing, mix thoroughly before using. Repeated freezing and thawing of the product is not recommended. However, the product demonstrated no loss of performance after 20 freeze-thaw cycles or 2 months at +20°C.
Instrument Compatibility:
Different real-time PCR systems employ different strategies for normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument. KiCqStart SYBR Green qPCR ReadyMix does not contain an internal reference dye. Compatible instruments include:
  • Bio-Rad® CFX384
  • Bio-Rad CFX96
  • Bio-Rad MiniOpticon
  • Bio-Rad/MJ Chromo4
  • Bio-Rad/MJ Opticon 2
  • Bio-Rad/MJ Opticon
  • Cepheid SmartCycler®
  • Eppendorf Mastercycler® ep realplex
  • Eppendorf Mastercycler ep realplex2 s
  • Illumina Eco qPCR
  • Qiagen/Corbett Rotor-Gene® 3000
  • Qiagen/Corbett Rotor-Gene 6000
  • Qiagen/Corbett Rotor-Gene Q
  • Roche LightCycler® 480

Legal Information

Bio-Rad is a registered trademark of Bio-Rad Laboratories, Inc.
CFX384 is a trademark of Bio-Rad Laboratories, Inc.
CFX96 is a trademark of Bio-Rad Laboratories, Inc.
Chromo4 is a trademark of Bio-Rad Laboratories, Inc.
Eppendorf is a registered trademark of Eppendorf AG
KiCqStart is a registered trademark of QIAGEN Beverly Inc.
LightCycler is a registered trademark of Roche
Mastercycler is a registered trademark of Eppendorf AG
MiniOpticon is a trademark of Bio-Rad Laboratories, Inc.
Opticon is a trademark of Bio-Rad Laboratories, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
Rotor-Gene is a registered trademark of Qiagen GmbH
SYBR is a registered trademark of Life Technologies
SmartCycler is a registered trademark of Cepheid, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phebe Verbrugghe et al.
BMC microbiology, 21(1), 23-23 (2021-01-13)
Since its discovery in 2007, the importance of the human gut bacterium Prevotella copri (P. copri) has been widely recognized with its links to diet and health status and potential as next generation probiotic. Therefore, precise, convenient and cost-effective diagnostic
NLRX1 Enhances Glutamate Uptake and Inhibits Glutamate Release by Astrocytes
Mahmoud S, et al.
Cells, 8(5), 400-400 (2019)
Muthappa Senthil-Kumar et al.
Nature protocols, 9(7), 1549-1562 (2014-06-06)
Tobacco rattle virus (TRV)-based virus-induced gene silencing (VIGS) is widely used in various plant species to downregulate the expression of a target plant gene. TRV is a bipartite, positive-strand RNA virus with the TRV1 and TRV2 genomes. To induce post-transcriptional
Tsion Zewdu Minas et al.
Oncotarget, 8(21), 34141-34163 (2016-05-19)
Ewing sarcoma (ES) involves a tumor-specific chromosomal translocation that produces the EWS-FLI1 protein, which is required for the growth of ES cells both in vitro and in vivo. However, an EWS-FLI1-driven transgenic mouse model is not currently available. Here, we
YK-4-279 effectively antagonizes EWS-FLI1 induced leukemia in a transgenic mouse model.
Minas TZ
Oncotarget, 6(35), 37678-37694 (2015)

Articles

After a traditional PCR has been completed, the PCR/qPCR data analysis is conducted by resolution through an agarose gel or, more recently, through a capillary.

PCR assay guide navigates you through primer validation and other assay optimization factors to ensure high sensitivity and specificity for optimum DNA/ RNA quantification.

Real-time polymerase chain reaction allows researchers to estimate the quantity of starting material in a sample. It has a much wider dynamic range of analysis than conventional PCR

Protocols

Quantitative PCR protocol using SYBR Green reagents. Procedure supports most qPCR instruments.

Quantitative PCR protocol using SYBR Green reagents. Procedure supports most qPCR instruments.

Quantitative PCR protocol using SYBR Green reagents. Procedure supports most qPCR instruments.

Quantitative PCR protocol using SYBR Green reagents. Procedure supports most qPCR instruments.

See All

Related Content

SYBR® Green I, a commonly used fluorescent DNA binding dye, binds all double-stranded DNA and detection is monitored by measuring the increase in fluorescence throughout the cycle. Explore our LuminoCt® and KiCqStart® products for Fast qPCR or JumpStart™ reagents for conventional qPCR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service