Skip to Content
Merck
All Photos(2)

Key Documents

KCQS07

Sigma-Aldrich

KiCqStart® One-Step Probe RT-qPCR ReadyMix

for Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems

Synonym(s):

KiCqStart® One Step Probe RT-qPCR Readymix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

form

liquid

usage

sufficient for 500 reactions

feature

dNTPs included
hotstart

storage condition

protect from light

technique(s)

qPCR: suitable

color

light blue

input

purified RNA

compatibility

Bio-Rad CFX384
Bio-Rad CFX96
Bio-Rad MJ Chromo4
Bio-Rad MJ Opticon 2
Bio-Rad MJ Opticon
Bio-Rad MiniOpticon
Eppendorf® Mastercycler ep realplex2 s
Eppendorf® Mastercycler ep realplex
Illumina Eco qPCR
Qiagen Corbett Rotor-Gene 3000
Qiagen Corbett Rotor-Gene 6000
Qiagen Corbett Rotor-Gene Q
Roche LightCycler 480

detection method

probe-based

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KiCqStart® One-Step Probe RT-qPCR ReadyMix is a ready-to-use, highly sensitive master mix for reverse transcription quantitative PCR (RT-qPCR) of RNA templates using hybridization probe detection chemistries such as TaqMan® 5′-hydrolysis probes on real-time PCR systems (refer to instrument compatibility section to select the appropriate reagent for your machine). First-strand cDNA synthesis and PCR amplification are carried out in the same tube without opening between procedures. It is ideal for highly sensitive quantification of RNA viruses or low abundance RNA targets as well as high throughput gene-expression studies. The system has been optimized to deliver maximum RT-PCR efficiency, sensitivity, and specificity in reduced reaction volumes and fast cycle times.

KiCqStart® One-Step Probe RT-qPCR ReadyMix contains all required components for RT-qPCR except RNA template and probe. It is compatible with all dual-labeled probe chemistries. The reverse transcriptase is an engineered M-MLV with reduced RNase H activity and improved activity and stability at higher temperatures. The use of higher temperatures (50 to 55°C) for the first-strand step of one-step RT-qPCR provides higher specificity for primer annealing and disruption of RNA secondary structure that can interfere with cDNA synthesis.

KiCqStart® One-Step Probe RT-qPCR ReadyMix is highly resistant to PCR inhibitors. A key component of the ReadyMix is an ultra-pure, highly processive, thermostable DNA polymerase that is combined with high avidity monoclonal antibodies. This provides an extremely stringent automatic hot-start that minimizes the potential for primer-dimer and other non-specific PCR artifacts. The light blue color of the inert dye simplifies reaction assembly in white, or clear, plates and helps to minimize pipetting or mixing errors. The dye does not interfere with qPCR performance or affect the stability of the product.

Application

KiCqStart® One-Step Probe RT-qPCR ReadyMix has been used:
  • in quantitative real-time polymerase chain reaction (qRT-PCR) for gene expression studies
  • in cell type-specific transcript analysis for determining the cycle threshold (Ct) value
  • for severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) RNA detection from nasopharyngealand oropharyngeal swabs
  • for amplification of reverse-transcribed cDNA

Features and Benefits

  • Assay results in as little as 33 minutes
  • Highly efficient and sensitive real-time PCR results
  • Little/no optimization required

Components

2X reaction buffer containing dATP, dCTP, dGTP, dTTP, magnesium chloride, reverse transcriptase, RNase inhibitor protein, hot-start DNA polymerase, inert blue qPCR dye, and stabilizers

packaging:
500 reactions* = 5 X 1 mL tubes
*number of reactions based on a 20uL volume

Other Notes

Different real-time PCR systems employ different strategies for normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument.KiCqStart® One-Step Probe RT-qPCR ReadyMix does not contain an internal reference dye. Compatible instruments include:
  • Bio-Rad® CFX384
  • Bio-Rad CFX96
  • Bio-Rad MiniOpticon
  • Bio-Rad/MJ Chromo4
  • Bio-Rad/MJ Opticon 2
  • Bio-Rad/MJ Opticon
  • Cepheid SmartCycler®
  • Eppendorf Mastercycler® ep realplex
  • Eppendorf Mastercycler ep realplex2 s
  • Illumina Eco qPCR
  • Qiagen/Corbett Rotor-Gene® 3000
  • Qiagen/Corbett Rotor-Gene 6000• Qiagen/Corbett Rotor-Gene Q
  • Roche LightCycler® 480

Legal Information

Bio-Rad is a registered trademark of Bio-Rad Laboratories, Inc.
CFX384 is a trademark of Bio-Rad Laboratories, Inc.
CFX96 is a trademark of Bio-Rad Laboratories, Inc.
Chromo4 is a trademark of Bio-Rad Laboratories, Inc.
Eppendorf is a registered trademark of Eppendorf AG
KiCqStart is a registered trademark of QIAGEN Beverly Inc.
LightCycler is a registered trademark of Roche
Mastercycler is a registered trademark of Eppendorf AG
MiniOpticon is a trademark of Bio-Rad Laboratories, Inc.
Opticon is a trademark of Bio-Rad Laboratories, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
Rotor-Gene is a registered trademark of Qiagen GmbH
SmartCycler is a registered trademark of Cepheid, Inc.
TaqMan is a registered trademark of Roche Molecular Systems, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin-resistant subjects have normal angiogenic response to aerobic exercise training in skeletal muscle, but not in adipose tissue
Walton R G, et al.
Physiological Reports, 3(6), 145-153 (2015)
Myonuclear transcription is responsive to mechanical load and DNA content but uncoupled from cell size during hypertrophy
Kirby T J, et al.
Molecular Biology of the Cell, 27(5), 788-798 (2016)
Bhasker Dharavath et al.
Heliyon, 6(7), e04405-e04405 (2020-07-16)
Early diagnosis of SARS-CoV-2 infected patients is essential to control the dynamics of the COVID-19 pandemic. We develop a rapid and accurate one-step multiplex TaqMan probe-based real-time RT-PCR assay, along with a computational tool to systematically analyse the data. Our
Fabio Sossai Possebon et al.
Journal of virological methods, 300, 114414-114414 (2021-12-14)
COVID-19 has posed a worldwide public health challenge affecting millions of people in different countries. Rapid and efficient detection of SARS-CoV-2 is essential for pandemic control. Reverse Transcription quantitative PCR (RT-qPCR) of nasopharyngeal swabs is the gold standard method for
Integrative mRNA-microRNA analyses reveal novel interactions related to insulin sensitivity in human adipose tissue
Kirby T J, et al.
Physiological Genomics, 48(2), 145-153 (2015)

Articles

Viral RNA Extraction Buffer (VRE100) and related reagents for qRT-PCR and additional viral RNA analyses.

RT-qPCR products combine the effective of Reverse Transcriptase with hot-start taq-directed antibody in convenient ReadyMixes for probe-based or SYBR® Green based applications.

PCR assay guide navigates you through primer validation and other assay optimization factors to ensure high sensitivity and specificity for optimum DNA/ RNA quantification.

Challenges in gene expression analysis include mRNA stability, temporal transcription patterns, and mRNA-protein correlation, impacting accuracy.

See All

Protocols

KiCqStart One-Step Probe RT-qPCR ReadyMix streamlines RNA template RT-qPCR with high sensitivity.

KiCqStart One-Step Probe RT-qPCR ReadyMix streamlines RNA template RT-qPCR with high sensitivity.

KiCqStart One-Step Probe RT-qPCR ReadyMix streamlines RNA template RT-qPCR with high sensitivity.

KiCqStart One-Step Probe RT-qPCR ReadyMix streamlines RNA template RT-qPCR with high sensitivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service