Skip to Content
Merck
All Photos(1)

Key Documents

I9783

Sigma-Aldrich

Monoclonal Anti-Importin α3 antibody produced in rat

clone 3D10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Imp α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

mouse, hamster, bovine, canine, rat, human, monkey

technique(s)

microarray: suitable
western blot: 2-4 μg/mL using HeLa cell extract

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KPNA4(3840)
mouse ... Kpna4(16649)
rat ... Kpna4(361959)

Immunogen

recombinant mouse importin α3/Qip1.

Biochem/physiol Actions

The gene KPNA4 (karyopherin subunit α 4) encodes a 16kDa protein, also referred to as importin α 3 belonging to the family of karyopherins (designated KPNA1 to KPNA4), that participates in the recognition of NLS (nuclear localization signal) sites and dock NLS-containing proteins destined for transport into the nuclear pore complex. It binds to translocon and participates in the sorting of INM (inner nuclear membrane)-directed integral membrane proteins. It associates with nucleoporins and functions in cargo-translocation across the nuclear membrane.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

KPNA4 Mediates Vitamin D-Dependent Inhibition of NF-kB Activity in Swine Epicardial Preadipocytes.
Chen S
Circulation, A12818-A12818 (2014)
Matthew Frieman et al.
Journal of virology, 81(18), 9812-9824 (2007-06-29)
The host innate immune response is an important deterrent of severe viral infection in humans and animals. Nuclear import factors function as key gatekeepers that regulate the transport of innate immune regulatory cargo to the nucleus of cells to activate
Sharon C Braunagel et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(22), 9307-9312 (2007-05-23)
Spodoptera frugiperda (Sf9) importin-alpha-16 is a translocon-associated protein that participates in the early sorting pathway of baculovirus integral membrane proteins destined for the inner nuclear membrane (INM). To discern whether sorting intermediate protein complexes like those observed in insect cells
Annett Koch et al.
Cell cycle (Georgetown, Tex.), 10(8), 1200-1207 (2011-03-18)
It is well established that mutual relations exist between the oxidative status of a cell and the ubiquitin proteasome system (UPS). Oxidation of proteins leads to misfolding and thus claims a higher proteolytic activity of the UPS to dispose these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service