I9658
Anti-Importin α1 antibody, Rat monoclonal
clone 1A6, purified from hybridoma cell culture
Synonym(s):
Anti-Imp α, Monoclonal Anti-Importin α1 antibody produced in rat
About This Item
Recommended Products
biological source
rat
conjugate
unconjugated
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
clone
1A6, monoclonal
form
buffered aqueous solution
mol wt
antigen ~60 kDa
species reactivity
human
technique(s)
immunocytochemistry: suitable
microarray: suitable
western blot: 0.1-0.2 μg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... KPNA2(3838)
mouse ... Kpna2(16647)
General description
Specificity
Immunogen
Application
- immunoblotting
- immunofluorescence
- immunocytochemistry
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service