Skip to Content
Merck
All Photos(3)

Key Documents

I8000

Sigma-Aldrich

2-Iodosobenzoic acid

≥98% (TLC)

Synonym(s):

IBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OIC6H4CO2H
CAS Number:
Molecular Weight:
264.02
Beilstein:
1939973
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

2-Iodosobenzoic acid,

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

230 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

OC(=O)c1ccccc1[I]=O

InChI

1S/C7H5IO3/c9-7(10)5-3-1-2-4-6(5)8-11/h1-4H,(H,9,10)

InChI key

IFPHDUVGLXEIOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

For cleavage of tryptophanyl peptide bonds.

Biochem/physiol Actions

2-Iodosobenzoic acid (IBA) may be used to study enzyme structure and activity. IBA is used as an oxidant which oxidizes vicinal sulfhydryls to disulfides (cysteine residues) within enzymes leading to their inactivation or conformational changes. IBA is also used to cleave tryptophanyl peptide bonds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Fontecave et al.
Biochemical and biophysical research communications, 168(2), 659-664 (1990-04-30)
The active form of protein B2, the small subunit of ribonucleotide reductase from E. Coli, contains a binuclear non heme iron center and a tyrosyl radical. MetB2 is an inactive form that lacks the radical but retains the Fe(III) center.
J O Moskaug et al.
The Journal of biological chemistry, 266(4), 2652-2659 (1991-02-05)
When the enzymatically active A-fragment of diphtheria toxin is translocated to the cytosol, the B-fragment inserts into the membrane in such a way that a 25-kDa polypeptide becomes shielded from proteases added to the external medium. We have attempted to
M F Drincovich et al.
Biochimica et biophysica acta, 1206(1), 10-16 (1994-05-18)
Incubation of C4 NADP-malic enzyme from maize leaves with the oxidant o-iodosobenzoate leads to the reversible and complete inactivation of the enzyme. The time-course of inactivation is biphasic with the rate depending on the o-iodosobenzoate concentration. The inactivation is partially
Neal N Reed et al.
Bioorganic & medicinal chemistry letters, 12(15), 2047-2049 (2002-07-13)
A series of soluble and insoluble polymer supported versions of the versatile oxidizing reagent IBX has been prepared. Each of the reagents were evaluated for their efficiency in the conversion of benzyl alcohol to benzaldehyde. Results from this study were
A Magalhaes et al.
FEBS letters, 329(1-2), 116-120 (1993-08-23)
The complete amino acid sequence of a thrombin-like enzyme with gyroxin activity isolated from the venom of the bushmaster snake Lachesis muta muta was determined by automated and DABITC/PITC microsequencing of the intact protein; fragments derived from it by separate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service