Skip to Content
Merck
All Photos(4)

Key Documents

228036

Sigma-Aldrich

OXONE®, monopersulfate compound

Synonym(s):

Potassium peroxymonosulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHSO5 · 0.5KHSO4 · 0.5K2SO4
CAS Number:
Molecular Weight:
307.38
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

vapor pressure

<0.0000017 hPa

form

powder

reaction suitability

reagent type: oxidant

concentration

>4.0% (active oxygen basis (by Na2S2O3, titration))

pH

2.1 (77 °C, 30 g/L)

SMILES string

[K+].[K+].[K+].[K+].[K+].OS([O-])(=O)=O.[O-]S([O-])(=O)=O.O[S+]([O-])([O-])(=O)=O.O[S+]([O-])([O-])(=O)=O

InChI

1S/5K.2H2O5S.2H2O4S/c;;;;;2*1-5-6(2,3)4;2*1-5(2,3)4/h;;;;;2*1H,(H,2,3,4);2*(H2,1,2,3,4)/q5*+1;;;;/p-5

InChI key

HJKYXKSLRZKNSI-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

General description

OXONE®, monopersulfate compound is a potassium triple salt mainly used as a stable, easy to handle and nontoxic oxidant.[1]

Application

A rapid, efficient synthesis of oxaziridines from imines using buffered OXONE has been reported.[2] Also used to study fading of an artist′s colorants.[3]
OXONE®, monopersulfate compound may be used as an alternative to transition-metal oxidants for the conversion of aldehydes to carboxylic acids or esters.[1]
Oxidant used for halogenation of α,β-unsaturated carbonyl compounds and catalytic generation of hypervalent iodine reagents for alcohol oxidation.

Legal Information

OXONE is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yifu Li et al.
Bioresource technology, 262, 294-301 (2018-05-08)
In this study, zero valent iron (ZVI) activated peroxymonosulfate (PMS) as novel technique (i.e. ZVI-PMS technology) was employed to enhance sludge dewatering. In optimal sludge dewatering conditions of ZVI and KHSO5 dosages, the specific resistance to filtration (SRF) was reduced
Atmos. Environ., Part A, 27A, 765-765 (1993)
Yao-Hui Huang et al.
Journal of hazardous materials, 170(2-3), 1110-1118 (2009-06-23)
In order to generate powerful radicals as oxidizing species for the complete decolorization and degradation of azo dye Reactive Black B (RBB) at near neutral pH (pH 6), homogeneous activation of peroxymonosulfate (Oxone: PMS) by the trace Co2+-catalysts was explored.
Benjamin R Travis et al.
Organic letters, 5(7), 1031-1034 (2003-03-28)
[reaction: see text] A highly efficient, mild, and simple protocol is presented for the oxidation of aldehydes to carboxylic acids utilizing Oxone as the sole oxidant. Direct conversion of aldehydes in alcoholic solvents to their corresponding ester products is also
Yi-Fong Huang et al.
Journal of hazardous materials, 167(1-3), 418-426 (2009-02-14)
This study investigated the degradation and mineralization of Bisphenol A (BPA) at pH 7, taken as a model compound in the presence of the trace metal-ions, Co(2+), and peroxymonosulfate (Oxone: PMS). We took advantage of the high oxidation-reduction potential of

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. do you need explosive rated fans to operate around Oxone?

    1 answer
    1. The Safety Data Sheet classifies this material as non-explosive therefore there is no need for explosive rated fans in order to operate around OXONE®, monopersulfate compound Product 228036-100G.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service