Skip to Content
Merck
All Photos(1)

Documents

I6659

Sigma-Aldrich

Imperatorin

≥98% (HPLC), powder

Synonym(s):

8-Isopentenyloxypsoralene, 9-(3-Methylbut-2-enyloxy)-7H-furo[3,2-g]chromen-7-one, 9-[(3-Methyl-2-buten-1-yl)oxy]-7h-furo[3,2-g][1]benzopyran-7-one, Ammidin, Marmelide, Marmelosin, NSC 402949

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white to light brown

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

C\C(C)=C\COc1c2OC(=O)C=Cc2cc3ccoc13

InChI

1S/C16H14O4/c1-10(2)5-7-19-16-14-12(6-8-18-14)9-11-3-4-13(17)20-15(11)16/h3-6,8-9H,7H2,1-2H3

InChI key

OLOOJGVNMBJLLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Imperatorin is a modulator of p38, ERK pathway. Imperatorin increases BMP-2 expression (mRNA) and increases bone density/volume and mineralization in vivo.
Imperatorin is an important bioactive furanocoumarin. It is used to treat neurodegenerative diseases. It possesses anticancer, antibacterial, anti-inflammatory and HIV replication-inhibiting activities. Imperatorin serves as an inhibitor of acetylcholinesterase (AChE) and may help in treating Alzheimer′s disease (AD) and Parkinson′s disease (PD).

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yan Zhang et al.
Fitoterapia, 83(1), 60-66 (2011-10-11)
Augmented endothelial nitric oxide (NO) synthase (eNOS) signaling has been reported to be associated with improvements in cardiac remodeling, and NO levels have been shown to be related to cardiac hypertrophy and heart failure. Imperatorin, a dietary furanocoumarin, has been
Xia Lv et al.
Food chemistry, 138(4), 2260-2266 (2013-03-19)
Imperatorin (IMP) is a major constituent of many herbal medicines and possesses anti-osteoporosis activity. The present research work aimed to study the biotransformation processes of IMP and evaluated the anti-osteoporosis activity of the transformed metabolites. Among 18 strains of filamentous
Zhangqian Wang et al.
Pest management science, 68(7), 1041-1047 (2012-03-06)
An assessment was made of the toxicity of imperatorin and osthole identified in Cnidium monnieri fruit, 11 related compounds and five insecticides to larvae from insecticide-susceptible Culex pipiens pallens (KS-CP strain) and Aedes aegypti and wild C.p. pallens (YS-CP colony)
Ke-wang Luo et al.
Chemotherapy, 57(6), 449-459 (2011-12-23)
Imperatorin (IM) is a furanocoumarin isolated from the root of Angelica dahurica, which is reported to have anticonvulsant and anticancer effects. In this study, the antiproliferative effect of IM on 9 human cancer cell lines was examined, and human hepatoma
Barbara Budzynska et al.
Experimental and clinical psychopharmacology, 20(4), 325-332 (2012-06-13)
The purpose of the reported experiments was to examine the effects of imperatorin [9-(3-methylbut-2-enyloxy)-7H-furo[3,2-g]chromen-7-one], a bioactive furanocoumarin isolated from the fruits of Angelica archangelica (Angelica officinalis) on anxiety and memory-related behaviors of mice. Male Swiss mice were tested for anxiety

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service