Skip to Content
Merck
All Photos(1)

Documents

I2536

Sigma-Aldrich

ICI 192605

≥98% (HPLC)

Synonym(s):

(4Z)-rel-, 4(Z)-6-(2-o-chlorophenyl-4-o-hydroxyphenyl-1,3-dioxan-cis-5-yl) hexenoic acid, 4-Hexenoic acid, 6-[(2R,4R,5S)-2-(2-chlorophenyl)-4-(2-hydroxyphenyl)-1,3-dioxan-5-yl]-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23ClO5
CAS Number:
Molecular Weight:
402.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥20 mg/mL

originator

AstraZeneca

storage temp.

−20°C

SMILES string

OC(=O)CC\C=C/C[C@H]1CO[C@H](O[C@H]1c2ccccc2O)c3ccccc3Cl

InChI

1S/C22H23ClO5/c23-18-11-6-4-9-16(18)22-27-14-15(8-2-1-3-13-20(25)26)21(28-22)17-10-5-7-12-19(17)24/h1-2,4-7,9-12,15,21-22,24H,3,8,13-14H2,(H,25,26)/b2-1-/t15-,21+,22+/m0/s1

InChI key

WHUIENZXNGAHQI-YGPRPMEGSA-N

Biochem/physiol Actions

ICI 192605 is a potent thromboxane A2 receptor antagonist. It inhibits platelet aggregation and can reverse the effects of vasoconstrictors such as TXA2 or PGD2. ICI 192605 can reverse vasoconstriction induced by inhibition of NO production by L-NEMA, which leads to an increase in TXA2 release.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Manuel Daray et al.
British journal of pharmacology, 139(8), 1409-1416 (2003-08-19)
1. This study was undertaken to characterize pharmacologically the prostanoid receptor subtypes mediating contraction in human umbilical vein (HUV). 2. HUV rings were mounted in organ baths and concentration-response curves to U-46619 (TXA(2) mimetic) were constructed in the absence or
L A Shaw et al.
British journal of pharmacology, 117(5), 817-822 (1996-03-01)
1. The effects of the 5-HT2 antagonist, ICI 170,809 and the thromboxane A2 antagonist, ICI 192,605, given alone and in combination (n = 12 per group), were examined in anaesthetized rats. Haemodynamics and arrhythmias induced by permanent coronary artery occlusion
H Arakawa et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 24(6), 582-589 (1994-06-01)
Trimellitic anhydride (TMA) is a low molecular weight chemical which can cause occupational asthma. We studied the role of eicosanoids in airway responses to TMA at different times after sensitization in actively sensitized guinea-pigs. Sensitization was performed by two intradermal
J J Descombes et al.
European journal of pharmacology, 243(2), 193-199 (1993-10-19)
The goal of the present study was to characterize the role of the endothelium in the 5-hydroxytryptamine (5-HT)-induced contraction of the rat basilar artery. Rat basilar artery segments were mounted in myographs to study their isometric tension development. 5-HT caused
N al Jarad et al.
British journal of clinical pharmacology, 37(1), 97-100 (1994-01-01)
Many prostanoids including are prostaglandin (PG) F2 alpha and PGD2 are potent bronchoconstrictor agents. There is evidence to suggest that airway thromboxane (TP) receptor may act as a common receptor for their bronchoconstrictor actions. We tested the hypothesis that inhaled

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service