Skip to Content
Merck
All Photos(4)

Documents

HPA045511

Sigma-Aldrich

Anti-MICU2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EF-Hand Domain Family, Member A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LAAAVAGAALAGAGAAWHHSRVSVAARDGSFTVSAQKNVEHGIIYIGKPSLRKQRFMQFSSLEHEGEYYMTPRDFLFSVMFEQMERKTSVKKLTKKDIEDTLSGIQT

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EFHA1(221154)

General description

MICU2 (mitochondrial calcium uptake 2) belongs to the uniporter complex. It is vigorously expressed in visceral organs. It is usually seen in the mitochondrial intermembrane space. It is located on chromosome 13q11-q12.

Immunogen

EF-hand domain family, member A1 recombinant protein epitope signature tag (PrEST)

Application

Anti-MICU2 has been used in western blotting.

Biochem/physiol Actions

MICU2 (mitochondrial calcium uptake 2) plays an important role in the modulation of the mitochondrial calcium uniporter.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81358

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MICU2, a paralog of MICU1, resides within the mitochondrial uniporter complex to regulate calcium handling.
Plovanich M, et al.
PLoS ONE, 8(2):e55785 (2013)
Whole Genome Sequencing identifies homozygous BRCA2 deletion guiding treatment in de-differentiated prostate cancer.
Purshouse K, et al.
Cold Spring Harbor molecular case studies (2017)
Critical reappraisal confirms that Mitofusin 2 is an endoplasmic reticulum?mitochondria tether.
Naon D, et al.
Proceedings of the National Academy of Sciences of the USA, 113(40), 11249-11254 (2016)
Santhanam Shanmughapriya et al.
Nature communications, 9(1), 3449-3449 (2018-08-31)
Although many factors contribute to cellular differentiation, the role of mitochondria Ca2+ dynamics during development remains unexplored. Because mammalian embryonic epiblasts reside in a hypoxic environment, we intended to understand whether mCa2+ and its transport machineries are regulated during hypoxia.
Kimberli J Kamer et al.
EMBO reports, 15(3), 299-307 (2014-02-08)
The mitochondrial uniporter is a selective Ca(2+) channel regulated by MICU1, an EF hand-containing protein in the organelle's intermembrane space. MICU1 physically associates with and is co-expressed with a paralog, MICU2. To clarify the function of MICU1 and its relationship

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service