Skip to Content
Merck
All Photos(9)

Key Documents

HPA023099

Sigma-Aldrich

Anti-CALB1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Calbindin, Anti-Calbindin D28, Anti-D-28K, Anti-Vitamin D-dependent calcium-binding protein, avian-type

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:5000-1:10000

immunogen sequence

QSSLITASQFFEIWLHFDADGSGYLEGKELQNLIQELQQARKKAGLELSPEMKTFVDQYGQRDDGKIGIVELAHVLPTEENFLLLFRCQQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALB1(793)

General description

CALB1 (calbindin 1) is a member of the EF-hand family of calcium-binding proteins. It is mainly present in the cerebellum. The gene is mapped to human chromosome 8q21.

Immunogen

Calbindin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CALB1 (calbindin 1) participates in the Ca2+-dependant activities occuring in CALB1-expressing neurons. It is associated with the activation and maintenance of long term depression (LTD). It provides neuroprotection in the presence of oxidative stress. In mice model, absence of CALB1 causes severe neurological and behavioral disorders. Low expression of CALB1 is also linked with the cognitive abnormalities observed with MELAS (mitochondrial encephalomyopathy, lactic acidosis, and stroke-like episodes).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76216

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giorgi Kuchukhidze et al.
Epilepsia, 56(8), 1207-1216 (2015-06-18)
Alterations in γ-aminobutyric acid (GABA)-ergic cortical neurons have been reported in focal cortical dysplasia (FCD)Ia/IIIa, a malformation of cortical development associated with drug-resistant epilepsy. We compared numbers of neurons containing calcium-binding proteins parvalbumin (PV), calbindin (CB), and calretinin (CR) and
Paolo Flace et al.
Anatomical record (Hoboken, N.J. : 2007), 297(7), 1306-1315 (2014-04-11)
Calbindin-D28k (CB) is a calcium-binding protein largely distributed in the cerebellum of various species of vertebrates. As regards the human cerebellar cortex, precise data on the distribution of CB have not yet been reported. Aim of the present work was
Flávia C Nery et al.
Neurology. Genetics, 5(5), e353-e353 (2019-09-14)
To determine changes in serum profiles and kidney tissues from patients with spinal muscular atrophy (SMA) type 1 compared with age- and sex-matched controls. In this cohort study, we investigated renal structure and function in infants and children with SMA
Valentina Emmanuele et al.
Journal of the neurological sciences, 317(1-2), 29-34 (2012-04-10)
Mitochondrial encephalomyopathy, lactic acidosis, and stroke-like episodes (MELAS) is a maternally inherited mitochondrial syndrome characterized by seizures, migrainous headaches, lactic acidosis, vomiting, and recurrent stroke-like episodes. Patients often suffer from cognitive dysfunction of unclear pathogenesis. In this study, we explored
K Morishima et al.
Journal of human genetics, 44(6), 414-415 (1999-11-26)
A polymorphic dinucleotide (CA) sequence was isolated from a BAC clone containing the human 27-kD calbindin (CALB1) gene at 8q21. This polymorphism will be a useful genetic marker to study genetic variations of the CALB1 gene.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service