Skip to Content
Merck
All Photos(5)

Documents

HPA011917

Sigma-Aldrich

Anti-IGSF8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD316 antigen, Anti-CD81 partner 3, Anti-EWI-2, Anti-Glu-Trp-Ile EWI motif-containing protein 2, Anti-Immunoglobulin superfamily member 8 precursor, Anti-KCT-4, Anti-Keratinocytes-associated transmembrane protein 4, Anti-LIR-D1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PDVLQVSAAPPGPRGRQAPTSPPRMTVHEGQELALGCLARTSTQKHTHLAVSFGRSVPEAPVGRSTLQEVVGIRSDLAVEAGAPYAERLAAGELRLGKEGTDRYRMVVGGAQAGDAGTYHCTAAEWIQDPDGSWAQIAEKRAVLAHVDVQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGSF8(93185)

General description

IGSF8 (Immunoglobulin superfamily, member 8) is a cell surface IgSF protein with molecular weight of 63kDa. This novel member of Ig superfamily is predominantly expressed in normal human brain, liver and to a lesser extent in the glioblastoma tumors and cell lines. It is expressed on the B, T and natural killer cells but absent in the monocytes, polynuclear cells or platelets.

Immunogen

Immunoglobulin superfamily member 8 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

IGSF8 (Immunoglobulin superfamily, member 8) is majorly involved in the organization of actin cytoskeleton. It connects tetraspanin-enriched microdomains to actin cytoskeleton by the molecular interaction of tetraspanin-tetraspanin. It also binds to α-actinin mediated by PIP2 (Phosphatidylinositol 4,5-bisphosphate) in the regulation of immune synapse formation and T cell activation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71569

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphanie Charrin et al.
The Biochemical journal, 373(Pt 2), 409-421 (2003-04-24)
Several tetraspanins bind directly to a few molecular partners to form primary complexes, which might assemble through tetraspanin-tetraspanin interactions to form a network of molecular interactions, the tetraspanin web. We have produced a monoclonal antibody directed to a 63 kDa
Tatiana V Kolesnikova et al.
Neoplasia (New York, N.Y.), 11(1), 77-86 (2008-12-25)
EWI-2, a cell surface IgSF protein, is highly expressed in normal human brain but is considerably diminished in glioblastoma tumors and cell lines. Moreover, loss of EWI-2 expression correlated with a shorter survival time in human glioma patients, suggesting that
Mónica Gordón-Alonso et al.
Journal of immunology (Baltimore, Md. : 1950), 189(2), 689-700 (2012-06-13)
EWI motif-containing protein 2 (EWI-2) is a member of the Ig superfamily that links tetraspanin-enriched microdomains to the actin cytoskeleton. We found that EWI-2 colocalizes with CD3 and CD81 at the central supramolecular activation cluster of the T cell immune
Viviana Neviani et al.
The FEBS journal, 287(24), 5323-5344 (2020-03-18)
Lipidation of transmembrane proteins regulates many cellular activities, including signal transduction, cell-cell communication, and membrane trafficking. However, how lipidation at different sites in a membrane protein affects structure and function remains elusive. Here, using native mass spectrometry we determined that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service