Merck
  • Site-specific functionality and tryptophan mimicry of lipidation in tetraspanin CD9.
Site-specific functionality and tryptophan mimicry of lipidation in tetraspanin CD9.
The FEBS journal (2020-03-18)
Viviana Neviani, Sjoerd van Deventer, Tobias P Wörner, Katerina T Xenaki, Michiel van de Waterbeemd, Remco N P Rodenburg, Inge M N Wortel, Jeroen K Kuiper, Sofie Huisman, Joke Granneman, Paul M P van Bergen En Henegouwen, Albert J R Heck, Annemiek B van Spriel, Piet Gros
ABSTRACT
Lipidation of transmembrane proteins regulates many cellular activities, including signal transduction, cell-cell communication, and membrane trafficking. However, how lipidation at different sites in a membrane protein affects structure and function remains elusive. Here, using native mass spectrometry we determined that wild-type human tetraspanins CD9 and CD81 exhibit nonstochastic distributions of bound acyl chains. We revealed CD9 lipidation at its three most frequent lipidated sites suffices for EWI-F binding, while cysteine-to-alanine CD9 mutations markedly reduced binding of EWI-F. EWI-F binding by CD9 was rescued by mutating all or, albeit to a lesser extent, only the three most frequently lipidated sites into tryptophans. These mutations did not affect the nanoscale distribution of CD9 in cell membranes, as shown by super-resolution microscopy using a CD9-specific nanobody. Thus, these data demonstrate site-specific, possibly conformation-dependent, functionality of lipidation in tetraspanin CD9 and identify tryptophan mimicry as a possible biochemical approach to study site-specific transmembrane-protein lipidation.
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Anti-IGSF8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Eicosapentaenoyl ethanolamide, 5 mg/mL in absolute ethanol, ≥98% (HPLC)