Skip to Content
Merck
All Photos(7)

Documents

HPA011191

Sigma-Aldrich

Anti-GDF15 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Growth/differentiation factor 15 precursor, Anti-MIC-1, Anti-Macrophage inhibitory cytokine 1, Anti-NAG-1, Anti-NRG-1, Anti-NSAID-regulated protein 1, Anti-Placental TGF-β, Anti-Placental bone morphogenetic protein, Anti-Prostate differentiation factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LSLAEASRASFPGPSELHSEDSRFRELRKRYEDLLTRLRANQSWEDSNTDLVPAPAVRILTPEVRLGSGGHLHLRISRAALPEGLPEASRLHRALFRLSPTASRSWDVTRPLRRQLSLARPQAPALHLRLSPPPSQSDQL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GDF15(9518)

General description

GDF15 (growth differentiation factor 15) is a part of the transforming growth factor-β (TGFβ) or bone morphogenetic protein (BMP) superfamily. This gene resides in human chromosomal locus 19p12.1-13.1, and contains two exons. The pro-protein is a dimer containing a disulfide linkage which undergoes cleavage at RXXR sequence, by furin-like protease. The mature protein is a C-terminal dimer composed of 112 amino acids.

Immunogen

Growth/differentiation factor 15 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-GDF15 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

GDF15 (growth differentiation factor 15) is released in response to stress such as, inflammation, oxidative stress and hypoxia, from multiple tissues, such as heart and liver. Studies show that it is responsible for the down-regulation of microRNAs of muscles, leading to intensive care unit-acquired weakness (ICUAW) and related muscle atrophy. It also controls the developmental processes underlying embryogenesis, osteogenesis, and hematopoiesis. It also regulates adipose tissue function, cartilage and bone synthesis and immune response. It is involved in tissue injury and repair processes. It is up-regulated in melanoma, thyroid, breast, pancreatic, gastrointestinal, prostate and colorectal cancer. It is involved in tumorigenesis, and abnormal expression is related to poor prognosis in various cancers. It is a biomarker for multiple epithelial cancers such as, renal cell carcinoma, adenocarcinoma of the esophagus, ovary and colon. In glioblastomas, it acts as a tumor suppressor gene and induces apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72494

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiguo Zhang et al.
PloS one, 9(4), e96283-e96283 (2014-04-25)
Nonsteroidal anti-inflammatory drug (NSAID) activated gene-1 (NAG-1) is a divergent member of the transforming growth factor-beta (TGF-β) superfamily. NAG-1 plays remarkable multifunctional roles in controlling diverse physiological and pathological processes including cancer. Like other TGF-β family members, NAG-1 can play
Xiuying Liu et al.
PloS one, 10(5), e0127518-e0127518 (2015-05-23)
Hepatocellular carcinoma (HCC) and liver cirrhosis are associated with high mortality worldwide. Currently, alpha-fetoprotein (AFP) is used as a standard serum marker for the detection of HCC, but its sensitivity and specificity are unsatisfactory, and optimal diagnostic markers for cirrhosis
U Wallin et al.
British journal of cancer, 104(10), 1619-1627 (2011-04-07)
Growth differentiation factor 15 (GDF15) belongs to the transforming growth factor beta superfamily and has been associated with activation of the p53 pathway in human cancer. The aim of this study was to assess the prognostic value of GDF15 in
Maiko Okamoto et al.
Pathobiology : journal of immunopathology, molecular and cellular biology, 87(2), 100-113 (2020-01-03)
Growth differentiation factor 15 (GDF15), which is derived from tumor-associated macrophages (TAM) and cancer cells, promotes progression of esophageal squamous cell carcinomas (ESCC). However, its role in the ESCC microenvironment remains unclear. Here, we examined the effects of GDF15 on
Takayuki Ishige et al.
Scientific reports, 6, 21681-21681 (2016-02-20)
Gastric cancer is classified into two subtypes, diffuse and intestinal. The diffuse-type gastric cancer (DGC) has poorer prognosis, and the molecular pathology is not yet fully understood. The purpose of this study was to identify functional secreted molecules involved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service