Skip to Content
Merck
All Photos(11)

Key Documents

HPA008057

Sigma-Aldrich

Anti-INA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-66 kDa neurofilament protein, Anti-Alpha-Inx, Anti-Alpha-internexin, Anti-NF-66, Anti-Neurofilament-66

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ALDIEIAAYRKLLEGEETRFSTSGLSISGLNPLPNPSYLLPPRILSATTSKVSSTGLSLKKEEEEEEASKVASKKTSQIGESFEEILEETVISTKKTEKSNIEETTISSQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INA(9118)

General description

INA (internexin neuronal intermediate filament protein, α) is also called as α-Internexin. The gene is mapped to human chromosome 10q24.33 and the encoded protein colocalizes with the neurofilament (NF) triplet proteins (NF-L, NF-M, and NF-H) in the CNS. It is capable of self-assembly to form homopolymers.

Immunogen

Alpha-internexin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

INA (internexin neuronal intermediate filament protein, α) gene encodes a type-IV neuronal intermediate filament that functions in the morphogenesis of neurons. It is present within the pathological inclusions of NIFID (Neuronal intermediate filament inclusion disease) along with heavy, medium, and light neurofilament subunits (NF-L, NF-M, and NF-H). The disease is characterized by frontotemporal dementia, pyramidal, and extrapyramidal signs. α-Internexin serves as a prognostic biomarker for pancreatic neuroendocrine tumor aggressiveness.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70159

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bei Liu et al.
The Journal of clinical endocrinology and metabolism, 99(5), E786-E795 (2014-02-04)
We aimed to test whether α-internexin could be a molecular biomarker of tumor aggressiveness and prognosis in pancreatic neuroendocrine tumors (PNETs). Using immunohistochemical staining and Western blot, we detected the expression of α-internexin in 350 tumors from 343 patients, of
Shraddha Pai et al.
Nature communications, 10(1), 2046-2046 (2019-05-06)
Impaired neuronal processes, including dopamine imbalance, are central to the pathogenesis of major psychosis, but the molecular origins are unclear. Here we perform a multi-omics study of neurons isolated from the prefrontal cortex in schizophrenia and bipolar disorder (n = 55 cases
Aidong Yuan et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 26(39), 10006-10019 (2006-09-29)
Alpha-internexin, a neuronal intermediate filament protein implicated in neurodegenerative disease, coexists with the neurofilament (NF) triplet proteins (NF-L, NF-M, and NF-H) but has an unknown function. The earlier peak expression of alpha-internexin than the triplet during brain development and its
Nigel J Cairns et al.
The American journal of pathology, 164(6), 2153-2161 (2004-05-27)
Neuronal intermediate filament (IF) inclusion disease (NIFID) is a novel neurological disease of early onset with a variable clinical phenotype including frontotemporal dementia, pyramidal, and extrapyramidal signs. Pathologically, in affected areas, there is neuronal loss, astrocytosis, and neuronal intracytoplasmic aggregates
Octavian Bucur et al.
Nature protocols, 15(5), 1649-1672 (2020-04-03)
In pathology, microscopy is an important tool for the analysis of human tissues, both for the scientific study of disease states and for diagnosis. However, the microscopes commonly used in pathology are limited in resolution by diffraction. Recently, we discovered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service