Skip to Content
Merck
All Photos(6)

Documents

HPA007884

Sigma-Aldrich

Anti-ABCA3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ABC-C transporter antibody produced in rabbit, Anti-ATP-binding cassette 3 antibody produced in rabbit, Anti-ATP-binding cassette sub-family A member 3 antibody produced in rabbit, Anti-ATP-binding cassette transporter 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MLRLTLGEYGRTVVPFSVPGTSQLGQQLSEHLKDALQAEGQEPREVLGDLEEFLIFRASVEGGGFNERCLVAASFRDVGERTVVNALFNNQAYHSPATALAVVDNLLFKLLCGPHASIVVSNFPQPRSALQAAKDQFNEG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCA3(21)

General description

ABCA3 (ATP-binding cassette, sub-family A, member3) gene encodes a member of the ABCA subclass of ATP-binding cassette (ABC) transporters. Its expression is restricted to lungs in the type II cells expressing surfactant protein A. Its expression is up-regulated by glucocorticoids just before birth.

Immunogen

ATP-binding cassette sub-family A member 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-ABCA3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ABCA3 (ATP-binding cassette, sub-family A, member3) gene encodes a member of the ABC1 subfamily and is mainly localized to the limiting membrane of the lamellar bodies in human lung. It functions in the transmembrane transport of lipid components of pulmonary surfactant. It is involved in the metabolism of pulmonary surfactant lipids and in the biogenesis of lamellar body. Mutations in this gene are associated with fatal surfactant deficiency in neonates resulting in lethal respiratory distress and interstitial lung disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86886

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias R Overbeck et al.
Oncology, 84(6), 362-370 (2013-05-22)
Patients with advanced-stage bronchial cancer benefit from systemic cytostatic therapy, in particular from regimens integrating cisplatin and taxanes. However, eventual disease progression leads to a fatal outcome in most cases, originating from tumor cells resisting chemotherapy. We here show that
Tobias R Overbeck et al.
Oncology, 93(4), 270-278 (2017-07-07)
ATP-binding cassette transport protein A3 (ABCA3) is expressed in non-small cell lung cancer (NSCLC). We hypothesize that high-level ABCA3 expression may have a negative prognostic impact in patients with NSCLC. In 89 patients with NSCLC and curative intended surgery, we
Benjamin J van Soldt et al.
Development (Cambridge, England), 146(9) (2019-04-05)
Although the Hippo-yes-associated protein (Yap) pathway has been implicated in lung development, the specific roles for Yap and its nucleocytoplasmic shuttling in the developing airway and alveolar compartments remain elusive. Moreover, conflicting results from expression studies and differences in the
Nobuhiro Ban et al.
The Journal of biological chemistry, 282(13), 9628-9634 (2007-02-03)
ABCA3 protein is expressed predominantly at the limiting membrane of the lamellar bodies in alveolar type II cells, and mutations in the ABCA3 gene cause lethal respiratory distress in newborn infants. To investigate the function of ABCA3 protein, we generated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service