Skip to Content
Merck
All Photos(3)

Key Documents

A2512

Sigma-Aldrich

Albumin from chicken egg white

lyophilized powder, ≥98% (agarose gel electrophoresis)

Synonym(s):

Albumin from hen egg white, Ovalbumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.61
Pricing and availability is not currently available.

biological source

chicken egg white

Quality Level

type

Grade VI

Assay

≥98% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

44287 Da by calculation

purified by

chromatography

concentration

4—20 mol/mol albumin

extent of labeling

4-20 mol mannose per mol ovalbumin protein

technique(s)

bioconjugation: suitable

solubility

H2O: soluble 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... OVAL(396058)

Looking for similar products? Visit Product Comparison Guide

General description

Chicken egg albumin is the major protein constituent of egg whites. Chicken egg albumin is a phosphorylated-glycoprotein. From the amino acid sequence, the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa.[1]
Ovalbumin makes 75% of the total egg white protein.[2]

Application

A phosphorylated glycoprotein which is useful as a marker (~45 kDa) for protein electrophoresis. Has been used to produce an animal model of asthma.
Ovalbumin has been used:
  • as a medium for cell incubation in EliCell assay for the detection of released cytokines from eosinophil[3]
  • to induce allergic inflammation and atopic dermatitis in experimental mice[4]
  • as a component in control mixture for validating tandem mass spectral analysis[5]
  • as a model protein antigen to evaluate the in vivo efficacy of a chitosan/interleukin (IL)-12 adjuvant system[6]

Biochem/physiol Actions

Albumin from chicken egg white serves as a natural adhesive.[7]
Differential scanning calorimetry indicates that chicken egg albumin denatures at 84 °C.[8] Ovalbumin can be used as a carrier protein to conjugate to synthetic peptides for use as an immunogen. It has the following amino acids: 20 Lys, 10 Tyr, 6 Cys, 14 Asp, and 33 Glu which make it suitable for conjugation.

Physical form

Essentially salt free

Preparation Note

A further purification of A 5503 to reduce mannose content.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

<BIG>Unger A, et al.</BIG>
Conservation of Wood Artifacts: A Handbook, 554-554 (2013)
In vivo efficacy of a chitosan/IL-12 adjuvant system for protein-based vaccines
<BIG>Heffernan MJ, et al.</BIG>
Biomaterials, 32, 926-932 (2011)
<BIG>Mielenz JR, et al.</BIG>
Biotechnology for Fuels and Chemicals: The Twenty-Eighth Symposium.c, 928-928 (2019)
PTP1B deficiency exacerbates inflammation and accelerates leukocyte trafficking in vivo
<BIG>Berdnikovs S, et al.</BIG>
Journal of Immunology, 188, 874-884 (2012)
Experimental protein mixture for validating tandem mass spectral analysis
<BIG>Keller A, et al.</BIG>
Omics : A Journal of Integrative Biology, 6, 207-212 (2002)

Questions

  1. How can we make 1 mm concentration of chicken egg albumin

    1 answer
    1. The molecular weight of this material is 44,287 daltons or g/mol. A 1 molar solution would be 44,287 grams per liter, a 100 mM solution would be 4,428 grams per liter, and so on. To prepare 100 ml of a 1 mM solution, 4.4287 g of the product will be needed.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service