Skip to Content
Merck
All Photos(10)

Key Documents

HPA006385

Sigma-Aldrich

Anti-UBTF antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NOR-90, Anti-UBF, Anti-UBF1, Anti-UBF2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

WKLLSQKEKDAYHKKCDQKKKDYEVELLRFLESLPEEEQQRVLGEEKMLNINKKQATSPASKKPAQEGGKGGSEKPKRPVSAMFIFSEEKRRQLQEERPELSESELTRLLARMWNDLSEKKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBTF(7343)

General description

UBTF (upstream binding transcription factor) is composed of six HMG box domains, which are lined up consecutively. It also contains a dimerization and a transactivation domain. It has two isoforms- UBF1 and UBF2. It is also called NOR (nucleolus organizer region)-90, and is a member of the NOR family, which is a family of ribosomal genes. It is highly expressed in human fetus. This gene is localized to human chromosome 17q21.3.

Immunogen

Nucleolar transcription factor 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-UBTF antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

UBTF (upstream binding transcription factor) or NOR-90 is a transcriptionally competent NOR as it is active during interphase. It serves as a scaffold to conform chromatin in a transcriptionally favorable condition. It is involved in the transcription of rDNA (ribosomal DNA), when it interacts with Pol I (RNA polymerase 7) enzyme. Along with SL1 (selectivity factor 1) complex, it binds to the enhancer region of rDNA. It acts as a negative regulator of Herpes simplex virus 1 (HSV-1) replication, where it suppresses the expression of viral DNA. It plays a role in tissue development and leukemia development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70888

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Wolf et al.
The Biochemical journal, 453(3), 357-370 (2013-05-22)
Jmjd6 (jumonji-domain-containing protein 6) is an Fe(II)- and 2OG (2-oxoglutarate)-dependent oxygenase that catalyses hydroxylation of lysine residues in proteins involved in pre-mRNA splicing. Jmjd6 plays an essential role in vertebrate embryonic development and has been shown to modulate alternative splicing
Chun-Ping Cui et al.
Zhongguo ying yong sheng li xue za zhi = Zhongguo yingyong shenglixue zazhi = Chinese journal of applied physiology, 20(1), 66-69 (2004-02-01)
From large-scale sequence of human fetal liver cDNA library, we have obtained a full-length cDNA from an EST after further sequencing. It has been demonstrated by the alignment comparison with data base available that it is a novel member of
Margarita Sobol et al.
Nucleus (Austin, Tex.), 4(6), 478-486 (2014-02-12)
To maintain growth and division, cells require a large-scale production of rRNAs which occurs in the nucleolus. Recently, we have shown the interaction of nucleolar phosphatidylinositol 4,5-bisphosphate (PIP2) with proteins involved in rRNA transcription and processing, namely RNA polymerase I
Markéta Kalmárová et al.
Journal of structural biology, 163(1), 40-44 (2008-05-27)
Ribosomal genes are organized in clusters termed Nucleolus Organizer Regions (NORs). Essential components of the RNA polymerase I transcription machinery, including Upstream Binding Factor (UBF), can be detected on some NORs during mitosis; these NORs, termed competent, are believed to
Nigel D Stow et al.
The Journal of general virology, 90(Pt 1), 69-73 (2008-12-18)
Previous reports have shown that adenovirus recruits nucleolar protein upstream-binding factor (UBF) into adenovirus DNA replication centres. Here, we report that despite having a different mode of viral DNA replication, herpes simplex virus type 1 (HSV-1) also recruits UBF into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service