Skip to Content
Merck
All Photos(6)

Key Documents

HPA006295

Sigma-Aldrich

Anti-SIRT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NAD-dependent deacetylase sirtuin-1 antibody produced in rabbit, Anti-SIR2-like protein 1 antibody produced in rabbit, Anti-hSIR2 antibody produced in rabbit, Anti-hSIRT1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

IVTLLDQAAKSNDDLDVSESKGCMEEKPQEVQTSRNVESIAEQMENPDLKNVGSSTGEKNERTSVAGTVRKCWPNRVAKEQISRRLDGNQYLFLPPNRYIFHGAEVYSDSEDDVLSSSSCGSNSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIRT1(23411)

Immunogen

NAD-dependent deacetylase sirtuin-1 recombinant protein epitope signature tag (PrEST)

Application

Anti-SIRT1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Sirtuin 1 is a protein encoded by the SIRT1 gene in humans. It is a class III histone deacetylase. It plays an important role in biological functions (such as lifespan, stress, and inflammation) through deacetylation of histone and nonhistone proteins. This gene is involved in the chondrogenic development of mesenchymal stem cells (MSCs). It may be associated with the progression of colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70465

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Godfrin-Valnet et al.
Medical science monitor basic research, 20, 142-145 (2014-09-18)
Sirtuin 1 (SIRT1) is a class III histone deacetylase that may play a critical role in several biological functions, including lifespan, stress, and inflammation. Our main objective was to evaluate SIRT1 activity in peripheral blood mononuclear cells (PBMCs) in patients
Akira Noguchi et al.
Cancer medicine, 3(6), 1553-1561 (2014-08-26)
Clinical trials of histone deacetylase (HDAC) inhibitors as antitumor therapy have been conducted for gastric cancer. Expression of SIRT1, a class III HDAC, is related to poor prognosis in some malignancies. We investigated the correlation between SIRT1 expression and progression
Kewei Jiang et al.
Chinese medical journal, 127(11), 2021-2024 (2014-06-04)
Sirtuin 1 (SIRT1) has been reported to have diverse roles in various biological processes through deacetylation of histone and nonhistone proteins. However, the correlations among SIRT1 protein expression, clinicopathological parameters, and survival of colorectal cancer patients remain unclear. SIRT1 protein
Veena Theendakara et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(3), 685-700 (2016-01-23)
A major unanswered question in biology and medicine is the mechanism by which the product of the apolipoprotein E ε4 allele, the lipid-binding protein apolipoprotein E4 (ApoE4), plays a pivotal role in processes as disparate as Alzheimer's disease (AD; in
Han Yan et al.
Molecular medicine reports, 17(2), 2535-2542 (2017-12-06)
In the present study, the role of Forkhead Box O1 (FOXO1) in glioblastoma (GBM) cell tumorigenesis was examined and the underlying mechanisms were investigated. Reverse transcription‑quantitative polymerase chain reaction and western blot analyses were used to analyze the expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service