Skip to Content
Merck
All Photos(11)

Key Documents

ZRB07131

Sigma-Aldrich

Anti-Sirt1 (Sir2) Antibody, clone 1G16, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

NAD-dependent protein deacetylase sirtuin-1, Regulatory protein SIR2 homolog 1, SIR2-like protein 1, SIR2alpha, Sir2, mSIR2a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G16, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 80.32 kDa
observed mol wt ~110 kDa

species reactivity

mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry: 1:100-1:200
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1G16 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects murine Sirt-1. It targets an epitope with in the CLOCK interacting region.

Immunogen

His-tagged recombinant fragment corresponding to the first 131 amino acids from the N-terminal region of murine NAD-dependent protein deacetylase sirtuin-1 (Sirt-1).

Application

Anti-Sirt1 (Sir2), clone 1G16, ZooMAb is a recombinant Rabbit Monoclonal antibody that detects NAD-dependent protein deacetylase sirtuin-1 (Sirt1) and has been tested for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:200 dilution from a representative lot detected Sirt1 (Sir2) in mouse testis tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Sirt1 (Sir2) in NIH/3T3 cells.

Target description

NAD-dependent protein deacetylase sirtuin-1 (UniProt: Q923E4; also known as Regulatory protein SIR2 homolog 1, SIR2-like protein 1, SIR2alpha, Sir2, mSIR2a, Sirt-1) is encoded by the Sirt1 (also known as Sir2l1) gene (Gene ID: 93759) in murine species. Sirt1 is a NAD-dependent protein deacetylase that is widely expressed and links transcriptional regulation directly to intracellular energetics. In liver, Sirt1 is expressed in a circadian manner with maximal and minimal levels reaching at around Zeitgeber time (ZT) 16 and ZT4, respectively and its deacetylase activity is also regulated in a circadian manner that peaks at ZT15. It participates in the coordination of several cellular functions such as cell cycle, response to DNA damage, metabolism, apoptosis, and autophagy. It is shown to modulate chromatin function through deacetylation of histones and can promote alterations in the methylation of histones and DNA, leading to transcriptional repression. It deacetylates a broad range of transcription factors and coregulators, thereby regulating target gene expression in both positive and negative manner. Sirt1 serves as a sensor of the cytosolic ratio of NAD+/NADH, which is altered by glucose deprivation and metabolic changes associated with caloric restriction. Sirt1 has four nucleotide (NAD)-binding regions and four zinc binding sites. It is reported to shuttle between nucleus and cytoplasm. Sir1 is proteolytically cleaved by cathepsin B upon TNF-alpha treatment to yield a catalytic inactive but stable 75 kDa fragment (aa 2-525). Sirt1 can be phosphorylated by multiple protein kinases. It is phosphorylated by STK4/MST1 that results in inhibition of SIRT1-mediated p53/TP53 deacetylation and phosphorylation by MAPK8/JNK1 at serine 46 and threonine 522 is reported to increase its nuclear localization and enzymatic activity. Sirt1 phosphorylation at threonine 522 by DYRK1A and DYRK3 activates its deacetylase activity and can promote cell survival. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service