Skip to Content
Merck
All Photos(7)

Key Documents

HPA001923

Sigma-Aldrich

Anti-SOX6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Transcription factor SOX-6 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TYKPGDNYPVQFIPSTMAAAAASGLSPLQLQQLYAAQLASMQVSPGAKMPSTPQPPNTAGTVSPTGIKNEKRGTSPVTQVKDEAAAQPLNLSSRPKTAEPVKSPTS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX6(55553)

Looking for similar products? Visit Product Comparison Guide

General description

Transcriptional factor SOX6 (SRY-box 6) is expressed in a wide variety of tissues, most abundantly in skeletal muscle and developing central nervous system (specifically in glioma tissues). It belongs to the Sry-related high-mobility group (HMG) box-containing gene family, that is involved in various developmental processes like neurogenesis, terminal differentiation and skeleton muscles formation. It has also reported that SOX6 gene is involved with human cardiac disorder.

Immunogen

Transcription factor SOX-6 recombinant protein epitope signature tag (PrEST)

Application

Anti-SOX6 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86388

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuko Hagiwara et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 234(2), 301-311 (2005-08-27)
We have previously demonstrated that p100H mutant mice, which lack a functional Sox6 gene, exhibit skeletal and cardiac muscle degeneration and develop cardiac conduction abnormalities soon after birth. To understand the role of Sox6 in skeletal muscle development, we identified
Ryo Ueda et al.
Oncogene, 23(7), 1420-1427 (2003-12-24)
To identify tumor antigens for glioma, a human testis cDNA library was screened by serological identification of antigens by recombinant expression cloning with sera from glioma patients. In this screening, the most frequently isolated antigen was SOX6, an Sry-related high-mobility
O Cohen-Barak et al.
Gene, 265(1-2), 157-164 (2001-03-20)
The Sox gene family encodes an important group of transcription factors harboring the conserved high-mobility group (HMG) box originally identified in the mouse and human testis determining gene Sry. We have cloned and sequenced SOX6, a member of the human
Tohid Siddiqui et al.
Methods in molecular biology (Clifton, N.J.), 2561, 159-170 (2022-11-19)
In this chapter, we present the methodology currently used in our laboratory to generate a starPEG-MMP (starPEG)- and heparin maleimide HM06 (heparin)-based 3D cell culture system, in a hydrogel, that can be used to study human neuronal development and Alzheimer's
R Fernández-Lloris et al.
FEBS letters, 580(5), 1215-1221 (2006-01-31)
SOX6 plays key functions in several developmental processes, including neurogenesis and skeleton formation. In this report, we show that SOX6 is modified in vitro and in vivo by small ubiquitin-related modifier (SUMO) on two distinct sites. Mutation of both sites

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service