Skip to Content
Merck
All Photos(8)

Key Documents

HPA001861

Sigma-Aldrich

Anti-STAT6 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

STAT Antibody - Anti-STAT6 antibody produced in rabbit, Stat6 Antibody, Anti-IL-4 Stat antibody produced in rabbit, Anti-Signal transducer and activator of transcription 6 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

VYPPHSHSIPPYQGLSPEESVNVLSAFQEPHLQMPPSLGQMSLPFDQPHPQGLLPCQPQEHAVSSPDPLLCSDVTMVEDSCLSQPVTAFPQGTWIGEDIFPPLLPPTEQDLTKLLLEGQGESGGGSLGAQPLLQPSHYGQSGISMSHMDLR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT6(6778)

General description

Signal transducers and activators of transcription 6 (STAT6) belongs to the family of DNA binding proteins. It is mainly involved with Interleukin-4 (IL-4)-mediated gene expression. Src homology-2 domain (SH2) domain of STAT6 binds to docking sites of IL-4 receptor α to get tyrosine-phosphorylated by the receptor-associated janus kinase (JAK). This binding activates SATAT6 and it forms homodimer. Further, it translocates to the nucleus for adhering with the high affinity STAT-binding elements (SBE) of IL-4/IL-13.

Immunogen

Signal transducer and activator of transcription 6 recombinant protein epitope signature tag (PrEST)

Application

Anti-STAT6 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77485

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hiroyuki Tateyama et al.
Immunology, 158(4), 340-352 (2019-09-15)
Siglecs are cell surface lectins that recognize sialic acids and are primarily expressed in hematopoietic cells. Previous studies showed that some Siglecs regulate macrophage function. In the present study, we examined the induction and putative roles of mouse Siglec-F in
H L Dickensheets et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(19), 10800-10805 (1999-09-15)
Interferons (IFNs) inhibit induction by IL-4 of multiple genes in human monocytes. However, the mechanism by which IFNs mediate this inhibition has not been defined. IL-4 activates gene expression by inducing tyrosine phosphorylation, homodimerization, and nuclear translocation of the latent
J Hou et al.
Science (New York, N.Y.), 265(5179), 1701-1706 (1994-09-16)
Interleukin-4 (IL-4) is an immunomodulatory cytokine secreted by activated T lymphocytes, basophils, and mast cells. It plays an important role in modulating the balance of T helper (Th) cell subsets, favoring expansion of the Th2 lineage relative to Th1. Imbalance
Hadjer Miloudi et al.
Cellular signalling, 46, 76-82 (2018-03-05)
Primary mediastinal B-cell lymphoma (PMBL) is a distinct B-cell lymphoma subtype with unique clinicopathological and molecular features. PMBL cells are characterised by several genetic abnormalities that conduct to the constitutive activation of the Janus kinase 2/signal transducer and activator of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service