Skip to Content
Merck
All Photos(7)

Documents

HPA001830

Sigma-Aldrich

Anti-SNAP25 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SNAP-25 antibody produced in rabbit, Anti-SUP antibody produced in rabbit, Anti-Super protein antibody produced in rabbit, Anti-Synaptosomal-associated 25 kDa protein antibody produced in rabbit, Anti-Synaptosomal-associated protein 25 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

SSDAYKKAWGNNQDGVVASQPARVVDEREQMAISGGFIRRVTNDARENEMDENLEQVSGIIGNLRHMALDMGNEIDTQNRQIDRIMEKADSNKTRIDEANQRATKMLGSG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNAP25(6616)

General description

Synaptosomal associated protein of 25kDa (SNAP25) is a nerve terminal protein with two isoforms, SNAP 25a and SNAP 25b. It is widely distributed in the frontal cortex and cerebellum of adult Down syndrome brain and plays an important role in Ca2+ dependent secretory vesicle-plasma membrane fusion in neuroendocrine cells. It is a component of a neuronal SNARE (soluble N-ethylmaleimide-sensitive factor attachment protein receptor) protein, essential for neurotransmitter release from presynaptic terminals. It binds directly to the vesicle-associated membrane protein such as syntaxin and synaptobrevin to mediate synaptobrevin binding for exocytosis.

Immunogen

Synaptosomal-associated protein 25 recombinant protein epitope signature tag (PrEST)

Application

Anti-SNAP25 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78868

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chueh-Ling Kuo et al.
PloS one, 6(5), e20352-e20352 (2011-06-02)
Botulinum neurotoxin (BoNT), a Category A biodefense agent, delivers a protease to motor neuron cytosol that cleaves one or more soluble NSF attachment protein receptors (SNARE) proteins involved in neurotransmission to cause a flaccid paralysis. No antidotes exist to reverse
Saori Yamamori et al.
The Journal of comparative neurology, 519(5), 916-932 (2011-02-01)
Soluble N-ethylmaleimide-sensitive factor attachment protein (SNAP)-25 is a neuronal SNARE protein essential for neurotransmitter release from presynaptic terminals. Three palmitoylated SNAP-25 family proteins: SNAP-25a, SNAP-25b, and SNAP-23, are expressed in the brain, but little is known about their distributions and
E R Chapman et al.
The Journal of biological chemistry, 269(44), 27427-27432 (1994-11-04)
The membrane proteins SNAP-25, syntaxin, and synaptobrevin (vesicle-associated membrane protein) have recently been implicated as central elements of an exocytotic membrane fusion complex in neurons. Here we report that SNAP-25 binds directly to both syntaxin and synaptobrevin. The SNAP-25-binding domain
I C Bark et al.
Gene, 139(2), 291-292 (1994-02-25)
Two distinct cDNA sequences, corresponding to alternative isoforms of the human nerve terminal protein SNAP-25 (synaptosomal associated protein of 25 kDa), were cloned and characterized. Sequence analysis demonstrated that the two isoforms are generated by alternative splicing between two distinct
Yoshikatsu Aikawa et al.
Molecular biology of the cell, 17(5), 2113-2124 (2006-02-17)
Soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) proteins play key roles in membrane fusion, but their sorting to specific membranes is poorly understood. Moreover, individual SNARE proteins can function in multiple membrane fusion events dependent upon their trafficking itinerary. Synaptosome-associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service