Skip to Content
Merck
All Photos(1)

Documents

H9148

Sigma-Aldrich

Hypercalcemia of malignancy factor fragment 1-34 amide human

≥97% (HPLC)

Synonym(s):

Ala-Val-Ser-Glu-His-Gln-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-NH2, PTHrP (1-34) amide, Parathyroid hormone-related protein (1-34) Amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C180H288N58O47
CAS Number:
Molecular Weight:
4016.57
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥97% (HPLC)

form

solid

technique(s)

drug transporter assay: suitable

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](Cc2c[nH]cn2)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc3ccccc3)NC(=O)[C@H](Cc4ccccc4)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@H](CCCCN)NC(=O)CNC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](Cc5c[nH]cn5)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](Cc6c[nH]cn6)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](C)N)C(C)C)[C@@H](C)CC)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](Cc7c[nH]cn7)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](C)C(N)=O

InChI

1S/C180H288N58O47/c1-23-94(16)141(174(282)208-99(21)147(255)210-116(51-55-137(247)248)156(264)235-143(96(18)25-3)176(284)231-128(72-107-79-197-87-206-107)170(278)238-144(100(22)241)177(285)207-98(20)145(186)253)236-169(277)121(65-92(12)13)222-164(272)125(69-104-76-194-84-203-104)228-165(273)126(70-105-77-195-85-204-105)226-160(268)120(64-91(10)11)220-161(269)123(67-102-41-30-27-31-42-102)224-162(270)122(66-101-39-28-26-29-40-101)223-152(260)112(47-38-60-200-180(191)192)213-150(258)110(45-36-58-198-178(187)188)212-151(259)111(46-37-59-199-179(189)190)214-157(265)117(61-88(4)5)221-168(276)130(74-139(251)252)229-154(262)114(49-53-134(185)243)217-175(283)142(95(17)24-2)237-172(280)132(82-240)232-149(257)109(44-33-35-57-182)209-135(244)80-201-148(256)108(43-32-34-56-181)211-167(275)129(73-138(249)250)230-166(274)127(71-106-78-196-86-205-106)227-159(267)119(63-90(8)9)219-158(266)118(62-89(6)7)218-153(261)113(48-52-133(184)242)215-163(271)124(68-103-75-193-83-202-103)225-155(263)115(50-54-136(245)246)216-171(279)131(81-239)233-173(281)140(93(14)15)234-146(254)97(19)183/h26-31,39-42,75-79,83-100,108-132,140-144,239-241H,23-25,32-38,43-74,80-82,181-183H2,1-22H3,(H2,184,242)(H2,185,243)(H2,186,253)(H,193,202)(H,194,203)(H,195,204)(H,196,205)(H,197,206)(H,201,256)(H,207,285)(H,208,282)(H,209,244)(H,210,255)(H,211,275)(H,212,259)(H,213,258)(H,214,265)(H,215,271)(H,216,279)(H,217,283)(H,218,261)(H,219,266)(H,220,269)(H,221,276)(H,222,272)(H,223,260)(H,224,270)(H,225,263)(H,226,268)(H,227,267)(H,228,273)(H,229,262)(H,230,274)(H,231,284)(H,232,257)(H,233,281)(H,234,254)(H,235,264)(H,236,277)(H,237,280)(H,238,278)(H,245,246)(H,247,248)(H,249,250)(H,251,252)(H4,187,188,198)(H4,189,190,199)(H4,191,192,200)/t94-,95-,96-,97-,98-,99-,100+,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,127-,128-,129-,130-,131-,132-,140-,141-,142-,143-,144-/m0/s1

InChI key

YUTJVTMXUNTCEG-WBTWNKCNSA-N

Gene Information

human ... PTHLH(5744)

Amino Acid Sequence

Ala-Val-Ser-Glu-His-Gln-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-NH2

Biochem/physiol Actions

Hypercalcemia of malignancy factor fragment 1-34 amide human is a parathyroid hormone-like protein (PLP) fragment used to study parathyroid hormone-related signaling pathways.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Kemp et al.
Science (New York, N.Y.), 238(4833), 1568-1570 (1987-12-11)
Peptides corresponding to the amino-terminal region of the parathyroid hormone-related protein (PTHrP) of humoral hypercalcemia of malignancy were synthesized. A 34-amino acid peptide, PTHrP(1-34), was two to four times more potent than bovine or human PTH(1-34) in bioassays promoting the
N Horiuchi et al.
Science (New York, N.Y.), 238(4833), 1566-1568 (1987-12-11)
One mechanism considered responsible for the hypercalcemia that frequently accompanies malignancy is secretion by the tumor of a circulating factor that alters calcium metabolism. The structure of a tumor-secreted peptide was recently determined and found to be partially homologous to
Koh Sato et al.
Seminars in oncology, 30(5 Suppl 16), 167-173 (2003-11-13)
Parathyroid hormone-related protein (PTHrP) plays a central role in humoral hypercalcemia of malignancy (HHM), which is one of the most frequent paraneoplastic syndromes. PTHrP produced by the tumor acts through a common PTH/PTHrP receptor to promote bone resorption, inhibit calcium
P Esbrit et al.
The Journal of endocrinology, 166(1), 11-20 (2000-06-17)
Parathyroid hormone (PTH)-related protein (PTHrP) is the main factor responsible for humoral hypercalcemia of malignancy. Both PTH and PTHrP bind to the common type I PTH/PTHrP receptor (PTHR), thereby activating phospholipase C and adenylate cyclase through various G proteins, in
H L Uy et al.
Cancer research, 57(15), 3194-3199 (1997-08-01)
Humoral hypercalcemia of malignancy results from the effects of tumor-produced factors on bone, kidney, and intestine that disrupt normal calcium homeostasis. Although parathyroid hormone-related protein (PTHrP) is a major mediator of the syndrome, tumors also produce other hypercalcemic factors, such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service