Skip to Content
Merck
All Photos(1)

Key Documents

10269611001

Roche

Neuraminidase (Sialidase)

from Arthrobacter ureafaciens

Synonym(s):

PCR, taq

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

bacterial (Arthrobacter ureafaciens)

Quality Level

form

solution

specific activity

~25 units/mg protein

packaging

pkg of 1 U (100 μl)

manufacturer/tradename

Roche

optimum pH

5.0-5.5

storage temp.

2-8°C

General description

Neuraminidase hydrolyzes terminal N- or O-acylneuraminic acids which are α2,3-, α2,6-, or α2,8-linked (rate: α2,6 > α2,3 > α2,8) to oligosaccharides, polysaccharides, mucopolysaccharides, glycoproteins, and glycolipids. Noteworthy, for the hydrolysis of glycolipids, the presence of a detergent is necessary. Because of the broad substrate specificity, the enzyme is very well suited for the complete removal of sialic acids from glycoconjugates of a wide variety of biological materials.[1]

Specificity

Cleaves terminal sialic-acid residues that are α2,3-, α2,6-, or α2,8-linked to Gal, GlcNAc, GalNAc, AcNeu, GlcNeu, oligosaccharides, glycolipids, or glycoproteins. Relative rate of cleavage is α2,6 >α2,3 >α2,8, determined on bonds in tri- and tetrasaccharides.

Application

  • cell surface lectin array analysis.[2][3][4]
  • hemagglutination assays.[5]
  • cell adhesion assay.[6]
For the hydrolysis of glycolipids, the presence of a detergent is necessary. Because of the broad substrate specificity, the enzyme is very well suited for the complete removal of sialic acids from glycoconjugates of a wide variety of biological materials.
Neuraminidase has been used for the:
  • detection of the cell surface glycosylations in human anaplastic large cell lymphoma cells[2]
  • release of sialic acid from cells[7]
  • antibody-overlay lectin microarray[4]

Physical properties

This product is a mixture of isoenzymes (L, M1, M2 and S) with the following molecular weight values: ~ 52 kDa, 66 kDa and 88 kDa.

Physical form

Solution in 10 mM sodium phosphate, 0.1% Micr-O-Protect (w/v), 0.25 mg/ml bovine serum albumin, pH 7

Preparation Note

Working concentration: Enzyme/substrate ratio should be in the range of 0.04 U/25-80 μg.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charting the Proteoform Landscape of Serum Proteins in Individual Donors by High-Resolution Native Mass Spectrometry.
Cramer, et al.
Analytical Chemistry, 94, 12732-12741 (2022)
Chao Gao et al.
bioRxiv : the preprint server for biology (2020-08-09)
The spike (S) glycoprotein in the envelope of SARS-CoV-2 is densely glycosylated but the functions of its glycosylation are unknown. Here we demonstrate that S is recognized in a glycan-dependent manner by multiple innate immune receptors including the mannose receptor
Cellular entry of the porcine epidemic diarrhea virus
Li W, et al.
Virus Research, 226, 117-127 (2016)
Tomonori Kaifu et al.
The Journal of experimental medicine, 218(12) (2021-11-25)
Dendritic cell immunoreceptor (DCIR) is a C-type lectin receptor with a carbohydrate recognition domain and an immunoreceptor tyrosine-based inhibitory motif. Previously, we showed that Dcir-/- mice spontaneously develop autoimmune enthesitis and sialadenitis, and also develop metabolic bone abnormalities. However, the
Sialylation by ??galactoside ??2,6?sialyltransferase and N?glycans regulate cell adhesion and invasion in human anaplastic large cell lymphoma
Suzuki O, et al.
International Journal of Oncology, 46(3), 973-980 (2015)

Protocols

Neuraminidase can be used to cleave sialic acids from proteins. In this protocol, the enzyme from Vibrio cholerae is used on fixed cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service