Skip to Content
Merck
All Photos(1)

Documents

H8889

Sigma-Aldrich

Histopaque®-1077 Hybri-Max

liquid, sterile-filtered, BioReagent, suitable for hybridoma

Synonym(s):

Histopaque 1077

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.75

sterility

sterile-filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | hybridoma: suitable
single cell analysis: suitable

impurities

endotoxin, tested

density

1.077 g/cm3 at 25 °C (lit.)

storage temp.

2-8°C

General description

Histopaque®-1077 is a solution comprising polysucrose and sodium diatrizoate, adjusted to a density of 1.077 +/- 0.001 g/cm3. This medium facilitates rapid recovery of viable lymphocytes from small volumes of whole blood. This technique is suitable for use in cell-mediated lympholysis and human lymphocyte antigen (HLA)2 typing. It may also be employed as the initial isolation step before the enumeration of T-, B-, and null lymphocytes. It is used to create a density medium for the purification of lymphocytes and other mononuclear cells.

Application

Histopaque®-1077 Hybri-Max has been used to isolate:
  • peripheral blood mononuclear cells (PBMCs) from heparinized blood by gradient centrifugation
  • immune cells (lymphocytes) from peripheral blood
  • PBMCs from blood

Legal Information

Histopaque is a registered trademark of Merck KGaA, Darmstadt, Germany
Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janet E Alexander et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 73(6), 720-728 (2017-11-11)
As in human populations, advances in nutrition and veterinary care have led to an increase in the lifespan of companion animals. Detrimental physiological changes occurring later in life must be understood before interventions can be made to slow or reduce
Mauricio S Caetano et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(24), 7576-7584 (2019-09-22)
Radiotherapy (RT) traditionally has been used for local tumor control in the treatment of cancer. The recent discovery that radiotherapy can have anticancer effects on the immune system has led to recognition of its ability to sensitize the tumor microenvironment
Siddharth Sharma et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 73(2), 160-167 (2008-01-15)
Coulter volume is far more accurate measure of cell volume than forward angle light scatter. In this report, we have used Coulter volume to determine the mean cell volume and diameter of normal human peripheral blood cells and hematopoietic progenitor
Joshua E Messinger et al.
mBio, 10(4) (2019-07-04)
Deciphering the molecular pathogenesis of virally induced cancers is challenging due, in part, to the heterogeneity of both viral gene expression and host gene expression. Epstein-Barr virus (EBV) is a ubiquitous herpesvirus prevalent in B-cell lymphomas of immune-suppressed individuals. EBV
Elliott D SoRelle et al.
eLife, 10 (2021-01-28)
Lymphoblastoid cell lines (LCLs) are generated by transforming primary B cells with Epstein-Barr virus (EBV) and are used extensively as model systems in viral oncology, immunology, and human genetics research. In this study, we characterized single-cell transcriptomic profiles of five

Protocols

This troubleshooting guide addresses the most common sources of error observed when using Histopaque but is not meant to be a comprehensive list.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service