Skip to Content
Merck
All Photos(2)

Documents

H4881

Sigma-Aldrich

Hydrocortisone 21-hemisuccinate sodium salt

≥90% (HPLC)

Synonym(s):

11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-hemisuccinate sodium salt, Cortisol 21-hemisuccinate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H33NaO8
CAS Number:
Molecular Weight:
484.51
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥90% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1CC[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(O)C(COC(CCC([O-])=O)=O)=O)=C1.[Na+]

InChI

1S/C25H34O8.Na/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30;/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30);/q;+1/p-1/t16-,17-,18-,22+,23-,24-,25-;/m0./s1

InChI key

HHZQLQREDATOBM-CODXZCKSSA-M

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Hydrocortisone 21-hemisuccinate sodium salt has been used:
  • as a component of the maturation medium for hepatic differentiation
  • in the corticosterone (CORT) treatment to study its effect on astrocyte-like neural stem cell proliferation in the area postrema of the adult mouse brain
  • in cortisol incubation experiments to study the effect of cortisol on the expression and function of sodium-chloride cotransporter (NCC) in zebrafish embryos

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuri Nambu et al.
Neuroscience letters, 748, 135732-135732 (2021-02-17)
The discovery of neural stem cells (NSCs) in the adult mammalian brain has provided insights into an extra level of brain plasticity. The proliferation and differentiation of NSCs is modulated by various physiological, pathological, and pharmacological stimuli. NSCs were recently
Chia-Hao Lin et al.
Molecular and cellular endocrinology, 422, 93-102 (2015-12-19)
In mammals, sodium/hydrogen exchanger (NHE) and sodium-chloride cotransporter (NCC) are expressed in renal tubules, and exhibit functional redundancy and mutual compensation in Na(+) uptake. In teleosts, the gills of the adult and skin of the embryonic stage function as external
Influence Of Cortisol On The Attachment And Metamorphosis Of Larval Utterbackia Imbecillis On Bluegill Sunfish (Lepomis Macrochirus).
Dubansky, B., et al.
The Biochemical Journal, 220, 97-106 (2011)
Yuri Nambu et al.
Neuroscience letters, 748, 135732-135732 (2021-02-17)
The discovery of neural stem cells (NSCs) in the adult mammalian brain has provided insights into an extra level of brain plasticity. The proliferation and differentiation of NSCs is modulated by various physiological, pathological, and pharmacological stimuli. NSCs were recently
Robert H Whitehead et al.
American journal of physiology. Gastrointestinal and liver physiology, 296(3), G455-G460 (2008-12-26)
It has proved to be impossible to culture epithelial cells from the gastrointestinal tract of adult animals. Researchers have had to use either cell lines derived from newborn rat small intestine or colon carcinoma cell lines that have retained some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service