Skip to Content
Merck
All Photos(1)

Key Documents

H4645

Sigma-Aldrich

Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin

≥90%

Synonym(s):

2,3,6-Tri-O-methyl-β-cyclodextrin, Trimethyl-β-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H112O35
CAS Number:
Molecular Weight:
1429.54
Beilstein:
78748
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90%

form

powder

technique(s)

electrophoresis: suitable

mp

170-178 °C (lit.)

storage temp.

2-8°C

SMILES string

COC[C@H]1O[C@@H]2O[C@@H]3[C@@H](COC)O[C@H](O[C@@H]4[C@@H](COC)O[C@H](O[C@@H]5[C@@H](COC)O[C@H](O[C@@H]6[C@@H](COC)O[C@H](O[C@@H]7[C@@H](COC)O[C@H](O[C@@H]8[C@@H](COC)O[C@H](O[C@H]1[C@H](OC)[C@H]2OC)[C@H](OC)[C@H]8OC)[C@H](OC)[C@H]7OC)[C@H](OC)[C@H]6OC)[C@H](OC)[C@H]5OC)[C@H](OC)[C@H]4OC)[C@H](OC)[C@H]3OC

InChI

1S/C63H112O35/c1-64-22-29-36-43(71-8)50(78-15)57(85-29)93-37-30(23-65-2)87-59(52(80-17)44(37)72-9)95-39-32(25-67-4)89-61(54(82-19)46(39)74-11)97-41-34(27-69-6)91-63(56(84-21)48(41)76-13)98-42-35(28-70-7)90-62(55(83-20)49(42)77-14)96-40-33(26-68-5)88-60(53(81-18)47(40)75-12)94-38-31(24-66-3)86-58(92-36)51(79-16)45(38)73-10/h29-63H,22-28H2,1-21H3/t29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43+,44+,45+,46+,47+,48+,49+,50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-/m1/s1

InChI key

DSDAICPXUXPBCC-MWDJDSKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin is a β-cyclodextrin derivative that finds potential use in pharmaceutical, food and cosmetic applications. Its attractive features include: the non-polar cavity desirable for its use as a molecular host for a wide range of molecules and its affinity for organic compounds in solution as well as crystalline phases.

Application

Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin may be used:
  • To investigate the crystal structure of its complexes with m-iodophenol, 4-biphenylacetic acid and (R)- and (S)-flurbiprofen by X-ray analysis.
  • To study the candidature of its complex with vitamin A for potential application as a drug delivery system for ophthalmic applications by high sensitivity fluorescence spectrometry and high pressure liquid chromatography (HPLC) techniques.
  • In the determination of the analyte composition in commercial samples by HPLC coupled to mass spectrometry (MS).

Chiral additive in liquid chromatography for enantiomer resolution. Chiral separation of basic drug substances by capillary electrophoresis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of Crystalline Complexes between Heptakis (2, 3, 6-tri-O-methyl)-beta-cyclodextrin and Various Alkanes or Alkenes (5? C? 10)?
Cardinael P, et al.
Journal of Inclusion Phenomena and Macrocyclic Chemistry, 39(1-2), 159-167 (2001)
Frantzeska Tsorteki et al.
Carbohydrate research, 339(2), 233-240 (2003-12-31)
The crystal structures of the complexes of heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin with indole-3-butyric acid and with 2,4-dichlorophenoxyacetic acid were studied by X-ray diffraction. The complexes crystallize in the monoclinic P2(1) space group. The host molecules are elliptically puckered and stacked along the a
I Bjørnsdottir et al.
Electrophoresis, 19(3), 455-460 (1998-04-29)
A capillary electrophoresis method for determination of the enantiomers of ibuprofen and its major phase I metabolites: 2'-hydroxyibuprofen and 2'-carboxyibuprofen in urine samples have been developed. Cyclodextrins and linear dextrins have been investigated as chiral selectors. Simultaneous chiral separation of
Juziro Nishijo et al.
Chemical & pharmaceutical bulletin, 52(12), 1405-1410 (2004-12-04)
The interaction of cholesterol with heptakis (2,3,6-tri-O-methyl)-beta-cyclodextrin (TOM-beta-CyD) was investigated in water using solubility method. It was found that TOM-beta-CyD forms two kinds of soluble complexes, with molar ratios of 1:1 and 1:2 (cholesterol:TOM-beta-CyD). The thermodynamic parameters for 1:1 and
Mingming Yin et al.
Se pu = Chinese journal of chromatography, 29(1), 91-94 (2011-05-18)
A new chiral stationary phase for gas chromatography, heptakis (2, 3, 6-tri-O-methoxymethyl)-beta-cyclodextrin, was synthesized by substituting the 2,3,6-OH groups of beta-cyclodextrin with methoxymethyl groups, and a coated capillary column for gas chromatography was made by coating this new stationary phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service