C4951
Cholesterol-Water Soluble
powder, BioReagent, suitable for cell culture
Synonym(s):
Cholesterol–methyl-β-cyclodextrin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352211
NACRES:
NA.75
Recommended Products
biological source
ovine wool
product line
BioReagent
form
powder
technique(s)
cell culture | mammalian: suitable
solubility
H2O: 300 mg/mL
shipped in
ambient
storage temp.
−20°C
Application
Cholesterol-Water Soluble has been used:
- in free cholesterol (FC) overloading of rat smooth muscle cells (SMCs)
- in cytotoxicity assay
- for the incubation of CD4+ T cells with Helicobacter pylori
- to study the membrane cholesterol level in SH-SY5Y human neuroblastoma cells
- to study the effects of cholesterol on cell viability and 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) reduction in primary cultured rat hippocampal neurons
- as a supplement for growth of bacterial culture
- to examine the interaction of S protein with angiotensin-converting enzyme 2 (ACE2)
- to investigate the effects of cholesterol on potassium currents in the inner hair cells of guinea pigs
- for reversible inhibition of proteinase K
Packaging
Package size based on cholesterol
Other Notes
Contains approx. 40 mg of cholesterol per gram; balance methyl-β-cyclodextrin.
Preparation Note
Preparea solution at 300mg/mL in water. Let thesolution sit undisturbed for one hour after diluting to obtain completesolubilization of product.
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Barbora Melkes et al.
Naunyn-Schmiedeberg's archives of pharmacology, 389(12), 1289-1300 (2016-09-08)
There are some indications that biased μ-opioid ligands may diversely affect μ-opioid receptor (MOR) properties. Here, we used confocal fluorescence recovery after photobleaching (FRAP) to study the regulation by different MOR agonists of receptor movement within the plasma membrane of
Cholesterol influences potassium currents in inner hair cells isolated from guinea pig cochlea
Kimitsuki T
Auris, Nasus, Larynx, 44(1), 46-51 (2017)
Cholesterol and its derivatives reversibly inhibit proteinase K
Singh N and Bhattacharyya D
Journal of Cellular Physiology, 232(3), 596-609 (2017)
Importance of cholesterol-rich membrane microdomains in the interaction of the S protein of SARS-coronavirus with the cellular receptor angiotensin-converting enzyme 2
Glende J, et al.
Virology, 381(2), 215-221 (2008)
A novel reciprocal and biphasic relationship between membrane cholesterol and beta-secretase activity in SH-SY5Y cells and in human platelets
Liu WW, et al.
Journal of Neurochemistry, 108(2), 341-349 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service