Skip to Content
Merck
All Photos(1)

Key Documents

18919-1AMP

Supelco

F.A.M.E. Mix, C4-C24

certified reference material,  wt. % (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

concentration

 wt. % (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat (solventless)

functional group

carboxylic acid
ester

shipped in

dry ice

storage temp.

−20°C

Related Categories

Application

It was used as calibration standard in determining fat content and FA distribution using GC/FID from berry seed press residues.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Analyte

Description

    cis-13,16-Docosadienoic acid methyl ester 2 wt. %
    cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester 2 wt. %
    cis-11,14-Eicosadienoic acid methyl ester 2 wt. %
    cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester 2 wt. %
    cis-8,11,14-Eicosatrienoic acid methyl ester 2 wt. %
    cis-11,14,17-Eicosatrienoic acid methyl ester 2 wt. %
    Methyl arachidate 4 wt. %
    Methyl arachidonate 2 wt. %
    Methyl behenate 4 wt. %
    Methyl butyrate 4 wt. %
    Methyl decanoate 4 wt. %
    Methyl cis-13-docosenoate 2 wt. %
    Methyl dodecanoate 4 wt. %
    Methyl cis-11-eicosenoate 2 wt. %
    Methyl elaidate 2 wt. %
    Methyl heneicosanoate 2 wt. %
    Methyl heptadecanoate 2 wt. %
    Methyl cis-10-heptadecenoate 2 wt. %
    Methyl hexanoate 4 wt. %
    Methyl linoleate 2 wt. %
    Methyl linolelaidate 2 wt. %
    Methyl linolenate 2 wt. %
    Methyl γ-linolenate 2 wt. %
    Methyl myristate 4 wt. %
    Methyl myristoleate 2 wt. %
    Methyl octanoate 4 wt. %
    Methyl oleate 4 wt. %
    Methyl palmitate 6 wt. %
    Methyl palmitoleate 2 wt. %
    Methyl pentadecanoate 2 wt. %
    Methyl cis-10-pentadecenoate 2 wt. %
    Methyl stearate 4 wt. %
    Methyl tetracosanoate 4 wt. %
    Methyl cis-15-tetracosenoate 2 wt. %
    Methyl tricosanoate 2 wt. %
    Methyl tridecanoate 2 wt. %
    Methyl undecanoate 2 wt. %
    See All (37)

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tom F O'Callaghan et al.
Journal of dairy science, 100(8), 6053-6073 (2017-06-19)
The purpose of this study was to investigate the effects of pasture-based versus indoor total mixed ration (TMR) feeding systems on the chemical composition, quality characteristics, and sensory properties of full-fat Cheddar cheeses. Fifty-four multiparous and primiparous Friesian cows were
Xu Liu et al.
Asian-Australasian journal of animal sciences, 33(12), 1957-1964 (2020-02-15)
The aim of this study was to compare overfeeding performance, fatty acid composition, blood chemistry, enzymes and genes expression overfed Xupu and Landes geese. Sixty male Xupu geese (80 d) and Landes geese (80 d) were selected. After a period
Capilla Mata-Pérez et al.
Frontiers in plant science, 6, 122-122 (2015-04-09)
Linolenic acid (Ln) released from chloroplast membrane galactolipids is a precursor of the phytohormone jasmonic acid (JA). The involvement of this hormone in different plant biological processes, such as responses to biotic stress conditions, has been extensively studied. However, the
Ru-Yi Li et al.
Journal of the science of food and agriculture, 98(3), 896-904 (2017-07-08)
Borage oil is a rich commercial source of γ-linolenic acid (18:3n-6). However, borage oil is rich in omega-6 polyunsaturated fatty acids and vulnerable to oxidation. Thus, selecting appropriate wall materials is critical to the encapsulation of borage oil. The present
M Sharifi et al.
Journal of animal physiology and animal nutrition, 102(1), 157-165 (2017-04-27)
This experiment was carried out to examine the effect of grapeseed oil (GSO) on performance, rumen fermentation, antioxidant status and subcutaneous adipose fatty acid (FA) profile in lambs. Eighteen Baluchi lambs, 196 ± 14 days of age and 39.8 ± 1.7 kg body weight, were randomly

Protocols

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

Chromatograms

suitable for GCsuitable for GCsuitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service